Informational:Inquire about OEM or Commercial Supply version of this product here.
Applied Biosystems™

GeneAmp™ 10X PCR Buffer II & MgCl2

Catalog number: N8080130
Have Questions?
Applied Biosystems™

GeneAmp™ 10X PCR Buffer II & MgCl2

Catalog number: N8080130
Have Questions?
Catalog Number
N8080130
Unit Size
6 x 1.5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 x 1.5 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 x 75 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
N80801306 x 1.5 mL
-
N80800101 x 1.5 mL
-
43798782 x 75 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Applied Biosystems™ GeneAmp™ 10X PCR Buffer II is optimized for use with AmpliTaq™ DNA Polymerase, resulting in robust PCR amplification. The provided MgCl2 solution offers versatility for optimizing PCR.

  • Optimized for use with AmpliTaq™ DNA Polymerase, offering maximum PCR performance
  • Available in several convenient sizes
  • Separate MgCl2 solution offers versatility

GeneAmp™ 10X PCR Buffer II, when diluted 1:10, provides the optimal pH and ionic strength for PCR amplification. GeneAmp™ 10X PCR Buffer II is magnesium ion free. A separate MgCl2 solution is provided. When used together, GeneAmp™ 10X PCR Buffer II and MgCl2 solution offer more versatility in optimizing the magnesium ion concentration to achieve optimal PCR amplification with any specific set of primers and DNA template.

Note: See user's manual or package insert for limited label license and trademark information.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase
AmpliTaq DNA Polymerase
Product Type
PCR Buffer
Quantity
6 x 1.5 mL
Shipping Condition
Dry Ice
For Use With (Application)
Standard PCR
Concentration
10X
Fidelity (vs. Taq)
1X
No. of Reactions
100 Reactions
Reaction Format
Kit
Volume
9 mL

Contents & Storage

• GeneAmp 10X PCR Buffer II, 6 x 1.5 mL
• MgCl2, 6 x 1.5 mL

Store at -15°C to -25°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text