Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
N8080155 | 5,000 units | 2,018.65 Online price 2,170.00 | - | ||
N8080160 | 250 units | 132.65 Online price 142.00 | - | ||
N8080171 | 1,000 units | 526.65 Online price 566.00 | - | ||
N8080152 | 3,000 units | 1,257.65 Online price 1,348.00 | - | ||
N8080185 | 25,000 units | 6,649.65 Online price 7,150.00 | - |
AmpliTaq DNA Polymerase is a 94 kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for the PCR process and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial, lot-to-lot.
Features of this enzyme:
Reliable and robust PCR
The thermal activity profile of AmpliTaq DNA Polymerase is good for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55°C to 75°C). The enzyme's half-life is ∼40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications. AmpliTaq DNA Polymerase is supplied with GeneAmp10X PCR Buffer I. It is also available with GeneAmp 10X PCR Buffer II and MgCl2 Solution.
For superior PCR performance, we recommend DreamTaq DNA Polymerase.