Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:Switch to Thermo Scientific DreamTaq DNA Polymerases for better results and added convenience.
Applied Biosystems™

AmpliTaq™ DNA Polymerase with Buffer II

Catalog number: N8080172
Have Questions?
Applied Biosystems™

AmpliTaq™ DNA Polymerase with Buffer II

Catalog number: N8080172
Have Questions?
Catalog Number
N8080172
Unit Size
1000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
250 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
3,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
5,000 units
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
25,000 units
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
N80801721000 units
-
N8080161250 units
-
N80801533,000 units
-
N80801565,000 units
-
N808018625,000 units
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

AmpliTaq DNA Polymerase is a 94 kDa, thermostable, recombinant DNA polymerase obtained by expression of a modified form of the Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is the most thoroughly characterized enzyme available for PCR and its recombinant nature and purification method provide unparalleled purity and reproducibility, vial-to-vial, lot-to-lot.

Features of this enzyme:

  • AmpliTaq DNA Polymerase is the most thoroughly characterized enzyme available for the PCR process, a testimony to its overall utility and efficacy
  • Its thermal activity profile makes it good for PCR applications
  • It is QC-tested to guarantee reproducible results

Reliable and robust PCR

The thermal activity profile of AmpliTaq DNA Polymerase is reliable for PCR applications because its optimal activity is in the same range at which stringent annealing of primers occurs (55–75°C). The enzyme's half-life is ∼40 minutes at 95°C, providing thermostability that meets the requirements of most difficult PCR applications. This AmpliTaqDNA Polymerase is supplied with GeneAmp 10X PCR Buffer II and MgCl2 Solution.

For superior PCR performance, we recommend DreamTaq DNA Polymerase.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Stand-alone enzyme
Polymerase
AmpliTaq DNA Polymerase
Reaction Speed
Standard
Exonuclease Activity
5' - 3'
Product Type
DNA Polymerase
Quantity
1000 Units
Shipping Condition
Dry Ice
For Use With (Application)
Standard PCR
Concentration
5 U/μL
Fidelity (vs. Taq)
1X
Hot Start
No
No. of Reactions
800 Reactions
Overhang
3'-A
Reaction Format
Separate Components
Size (Final Product)
5 kb or less
Starting Material
DNA

Contents & Storage

• AmpliTaq DNA Polymerase (5 U/μL), 200 μL
• GeneAmp 10X PCR Buffer II, 4 x 1.5 mL
• 25 mM MgCl2, 4 x 1.5 mL

Store at -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text