AmpliTaq Gold™ DNA Polymerase with Buffer II and MgCl2
Upgrade to Platinum II Taq Hot-Start DNA Polymerase for universal annealing and a 4X faster cycling protocol.
AmpliTaq Gold&trade; DNA Polymerase with Buffer II and MgCl<sub>2</sub>
Applied Biosystems™

AmpliTaq Gold™ DNA Polymerase with Buffer II and MgCl2

AmpliTaq Gold DNA Polymerase is a chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
N80802453,000 units
N8080241250 Units
N80802471,000 units
N808025925 x 1,000 units
N80802495,000 units
Catalog number N8080245
Price (USD)
3,050.65
Online Exclusive
3,280.00
Save 229.35 (7%)
Each
Add to cart
Quantity:
3,000 units
Request bulk or custom format
Price (USD)
3,050.65
Online Exclusive
3,280.00
Save 229.35 (7%)
Each
Add to cart
Ask our AI about this Product
AmpliTaq Gold DNA Polymerase with Buffer II and MgCl2 is a chemically modified form of AmpliTaq DNA Polymerase requiring thermal activation. The modified enzyme is provided in an inactive state and upon thermal activation, the modifier is permanently released, regenerating active enzyme. The resulting hot-start PCR amplification provides increased sensitivity, specificity, and yield over conventional PCR techniques.

AmpliTaq Gold DNA Polymerase can be activated partially or completely in a pre-PCR heat step or can be allowed to activate slowly in a time-released manner during the denaturation steps of thermal cycling. With or without a limited up-front heat activation step, active enzyme is released slowly during thermal cycling to match template concentration and increase specificity. The yield of specific product increases as reactants are not wasted in the formation of unintended products.

Features

  • Automated chemical hot-start enzyme for increased specificity, sensitivity, and yield
  • Time-released thermal activation improves sensitivity in low copy number amplifications
  • Successful multiplex PCR saves time and reagents
  • Includes GeneAmp 10X PCR Buffer II with MgCl2
  • Hot-start technology reduces risk of biological contamination

Notes

  • For better performance AmpliTaq Gold 360 DNA Polymerase is purified by an additional proprietary separation process to eliminate contaminating bacterial DNA sequences from the enzyme preparation.
  • When used with the enhanced 360 Gold Buffer, this ultra-pure enzyme reduces false positive results, amplifies low-level target sequences, and promotes the amplification of a variety of templates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration5 U/μL
Exonuclease Activity5' - 3'
Fidelity (vs. Taq)1X
FormatTube
Hot StartBuilt-In Hot Start
No. of Reactions2400 Reactions
Overhang3'-A
PolymeraseAmpliTaq Gold DNA Polymerase
Product TypeDNA Polymerase
Quantity3,000 units
Reaction FormatSeparate Components
Shipping ConditionDry Ice
Size (Final Product)5 kb or less
Starting MaterialDNA
For Use With (Application)Hot-start PCR
GC-Rich PCR PerformanceLow
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• AmpliTaq Gold DNA Polymerase (5 U/μL), 12 x 50 μL
• 10X PCR Buffer II, 12 x 1.5 mL
• 25 mM MgCl2, 12 x 1.5 mL

Store at -15°C to -30°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection