Search Thermo Fisher Scientific
Catalog Number | Description | Function | Includes |
---|---|---|---|
NATGAS020221 | Dual Channel NG & NGL Analyzer for GPA 2286, 2186 extended only | Both channels utilize a single Flame Ionization Detector (FID) to produce a detailed chromatogram and report for hydrocarbons from C1-C16. Provides The Extended Channel for Either or Both Liquid or Gas Samples According to GPA 2286 and 2186 Extended Analysis Only. |
|
NATGAS010011 | Single Channel NG Analyzer for GPA 2261 | Early C6+ Regroup Peak, Composite Peak for N2/Air, Individual Peaks for CO2, Methane, Ethane, and C3-C5 |
|
NATGAS010021 | Single Channel NG Analyzer for GPA 2261 O2/N2 | Early C6+ regroup peak, individual peaks for O2, N2, CO2, methane, ethane, C3-C5. Modified to Provide Separate Peaks for O2 and N2. |
|
NATGAS010031 | Single Channel NGL Analyzer for GPA 2177 | Early C7+ Regroup Peak, Composite Peak for N2/Air, Individual Peaks for CO2, Methane, Ethane, and C3-C6 |
|
NATGAS010211 | Single Channel NG C6+ or NGL C7+ Analyzer for GPA 2166 or 2177 | Early C6+ or C7+ Regroup Peak, Composite Peak for N2/Air, Individual Peaks for CO2, Methane, Ethane, and C3-C5 for Natural Gas or C3-C6 for Natural Gas Liquid |
|
NATGAS020011 | Dual Channel NG Analyzer for GPA 2261 | Each channel produces early C6+ regroup peak, composite peak for N2/air, individual peaks for CO2, methane, ethane, C3-C5. Allows Simultaneous Analysis of Two Different Samples. |
|
NATGAS020021 | Dual Channel NG Analyzer for GPA 2261 O2/N2 | Early C6+ regroup peak , individual peaks for O2, N2, CO2, methane, ethane, C3-C5 for each channel. Modified to Report Separate Peaks for O2 and N2. Allows Simultaneous Analysis of Two Different Samples. |
|
NATGAS020031 | Dual Channel NGL Analyzer for GPA 2177 | Early C7+ Regroup Peak, Composite Peak for N2/Air, Individual Peaks for CO2, Methane, Ethane, and C3-C6 |
|
NATGAS020061 | Dual Channel NG Analyzer for GPA 2286 | The two TCD channels produce early C6+ regroup peak, composite peak for N2/air, individual peaks for CO2, methane, ethane, and C3-C5. The two FID channels produce a detailed chromatogram and report for hydrocarbons from C1-C14 as required for the extended analysis in GPA 2286. Allows Simultaneous Analysis of Two Different Samples. |
|
NATGAS020211 | Dual Channel NG O2 N2 Analyzer & NGL for GPA 2261 2177 | Early C6+ regroup peak, individual peaks for O2, N2, CO2, methane, ethane, and C3-C5. The natural gas liquid channel produces early C7+ regroup peak. Modified to Provide Separate Peaks for O2 and N2. Allows Simultaneous Analysis of Two Samples (Gas, Liquid, or One of Each). |
|
NATGAS020231 | Dual Channel NG Analyzer for extended GPA 2286 | Channel One Produces Early C6+ Regroup Peak, Composite Peak for N2/Air, Individual Peaks for CO2, Methane, Ethane, and C3-C5 Channel Two Utilizes a Flame Ionization Detector (FID) to |
|
NATGAS020261 | Dual Channel NG Analyzer for GPA 2261 O2/N2 + GPA 2286 | Channel one produces early regroup peak C6+, individual peaks for O2, N2, CO2, methane, ethane, and C3-C5. Channel two utilizes a single Flame Ionization Detector (FID), one 6-port gas sampling valve located in the auxiliary oven, and a single capillary column located in the GC main oven. Channel two produces a detailed chromatogram and report for hydrocarbons from C1-C16. Modified to Report Separate Peaks for O2 and N2, or for GPA 2286 Extended Analysis of Natural Gas. |
|
NATGAS020271 | Single Channel NG Analyzer for GPA 2261 plus He/H2 | Early C6+ regroup peak , individual peaks for He, H2, air composite, CO2, methane, ethane, C3-C5. Has an Additional Channel for He/H2 Analysis. |
|
NATGAS020281 | Single Channel NG Analyzer for GPA 2261 O2/N2 and He/H2 | Early C6+ regroup peak, individual peaks for O2, N2, CO2, methane, ethane, C3-C5 and channel 2 TCD produces peaks for He and H2. Modified to Provide Separate Peaks for O2 and N2 with an Additional TCD Channel for Analysis of He/H2. |
|
NATGAS040011 | Four Channel NG Analyzer for GPA 2261 | Early C6+ Regroup Peak, Composite Peak for N2/Air, Individual Peaks for CO2, Methane, Ethane, and C3-C5 |
|
NATGASC20211 | Dual Channel NG Analyzer & NGL Analyzer for GPA 2261, 2177, 2286, 2186 | For Gas Samples, Channel One Produces Early C6+ Regroup Peak, Individual Peaks for O2, N2, CO2, Methane, Ethane, C3-C5 For Liquid Samples, Channel One Produces Early | TRACE 1310 GC with two independent channels
|
NATGASC20221 | Three Channel NG Analyzer He/H2 & NGL Analyzer for GPA 2261, 2177, 2186, 2286 | For Gas Samples, Channel One Produces Early C6+ Regroup Peak, Individual Peaks for O2, N2, CO2, Methane, Ethane, C3-C5 For Liquid Samples, Channel One Produces Early | TRACE 1310 GC with two independent channels
|
Select from over 15 turnkey Thermo Scientific™ TRACE™ 1310 Natural Gas and Natural Gas Liquid Analyzers for the vital information you need to meet increasing need for analytical data as natural gas (NG) and natural gas liquid (NGL) demand, production, custody transfer, and consumption. TRACE 1310 NG/NGL systems conduct analyses according to standard methods, GPA, ASTM, ISO, DIN, including calorific or BTU content, hydrocarbon speciation and impurities. Choose from low-cost single channel, best-value dual channel, or highly flexible combination systems.
Our team of specialist, with over 60 years in the business, has developed a complete line of analyzers. We offer over 15 different combinations to select from and all feature the innovative modularity of our Instant Connect injectors and detectors, providing the highest uptime and easiest maintenance.
Single channel analyzers – Lowest priced systems
Analyze a single sample per analysis using a single channel analyzer for NG or NGL.
Dual channel analyzers – Best value systems
Run two different samples simultaneously for improved productivity, or one sample with two channels of data BTU and speciation.
Combination analyzers – Most flexible system
A single platform capable of performing all the standard methods on a NG or NGL sample.
Share catalog number, name or link