Specifications
Absorbance Accuracy3% (at 0.74 Abs at 350 nm)
Absorbance Range0 to 75 A
Certifications/ComplianceUL/CSA and CE
CompatibilityProfessional versions of Microsoft™ Windows™ 7 (32 bit and 64 bit), Windows Vista (32 bit), and Windows XP (32 bit).
ConnectionsUSB
DescriptionNanoDrop 8000 Microvolume UV-Vis spectrophotometer
Detection Limits2.5 ng/μL (dsDNA), 0.15 mg/mL (BSA)
Detection Range2.5 to 3,700 ng/μLL (dsDNA), 0.15 - 100 mg/mL (BSA)
Footprint24 x 32 cm
For Use With (Application)Nucleic acid, protein, cell culture and custom methods
IncludesUSB cable and operating software
Measurement Time<20 sec.
ModelND-8000
No. of SamplesUp to 8
Pathlength0.2 and 1.0 mm
Power Consumption12 V DC
Sample Volume (Metric)1 μL
Spectral Resolution≤3 nm (FWHM at Hg 546.1 nm)
System RequirementsMicrosoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
Warranty1 year parts and labor
Wavelength Accuracy±1 nm
Weight (Metric)3.4 kg
Detector Type2048-element Linear Silicon CCD Array
Light SourceXenon Flash
TypeMicrovolume UV-Vis Spectrophotometer
Voltage12 VDC
Wavelength Range220 to 750 nm
Unit SizeEach