Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | NanoDrop OneC Productivity Kit |
For Use With (Equipment) | NanoDrop OneC |
Includes |
|
Storage Requirements | Store at room temperature |
Unit Size | Each |
Description | NanoDrop One Productivity Kit |
For Use With (Equipment) | NanoDrop One |
Includes |
|
Storage Requirements | Store at room temperature |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
ND-PP1C | Each | NanoDrop OneC Productivity Kit | 1,150.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
ND-PP1 | Each | NanoDrop One Productivity Kit | 829.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|