Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Absorbance Accuracy | 3% at 0.97 A, 302 nm, 23 ± 2°C |
Absorbance Range | 0.04 to 200 A (10 mm equivalent) |
Description | NanoDrop Eight Spectrophotometer F1 ClipTip Bundle |
Detection Limits | 2.0 to 10,000 ng/μL dsDNA; 0.03 to 145 mg/mL IgG |
Detector Type | 2048-element CMOS Linear Image Sensor |
Dimensions (L x W x H) | 24 x 33 x 17 cm (9.4 x 13 x 6.7 in.) |
Light Source | Xenon Flash |
Product Line | Microvolume |
Type | UV-Vis Spectrophotometer |
Voltage | 100/240 V |
Wavelength Range | 190 to 850 nm |
Measurement Time | 8 samples in ≤ 20 sec |
Pathlength | 1.0 mm, 0.2 mm, 0.1 mm (automatically adjusting) |
Power Consumption | 3 W at Idle; 15 W at Working Conditions |
Sample Volume (Metric) | 1 to 2 μL |
Spectral Resolution | ≤1.8 nm (FWHM at Hg 254 nm) |
Warranty | 1 Year |
Wavelength Accuracy | ± 1 nm |
Weight (English) | 8.2 lb. |
Weight (Metric) | 3.7 kg |
Unit Size | Each |
Absorbance Accuracy | 3% at 0.97 A, 302 nm, 23 ± 2°C |
Absorbance Range | 0.04 to 200 A (10 mm equivalent) |
Description | NanoDrop Eight Spectrophotometer E1 ClipTip Bundle |
Detection Limits | 2.0 to 10,000 ng/μL dsDNA; 0.03 to 145 mg/mL IgG |
Detector Type | 2048-element CMOS Linear Image Sensor |
Dimensions (L x W x H) | 24 x 33 x 17 cm (9.4 x 13 x 6.7 in.) |
Light Source | Xenon Flash |
Product Line | Microvolume |
Type | UV-Vis Spectrophotometer |
Voltage | 100/240 V |
Wavelength Range | 190 to 850 nm |
Measurement Time | 8 samples in ≤ 20 sec |
Pathlength | 1.0 mm, 0.2 mm, 0.1 mm (automatically adjusting) |
Power Consumption | 3 W at Idle; 15 W at Working Conditions |
Sample Volume (Metric) | 1 to 2 μL |
Spectral Resolution | ≤1.8 nm (FWHM at Hg 254 nm) |
Warranty | 1 Year |
Wavelength Accuracy | ± 1 nm |
Weight (English) | 8.2 lb. |
Weight (Metric) | 3.7 kg |
Unit Size | Each |
Absorbance Accuracy | 3% at 0.97 A, 302 nm, 23 ± 2°C |
Absorbance Range | 0.04 to 200 A (10 mm equivalent) |
Description | NanoDrop Eight Spectrophotometer with Laptop US/CAN |
Detection Limits | 2.0 to 10,000 ng/μL dsDNA; 0.03 to 145 mg/mL IgG |
Detector Type | 2048-element CMOS Linear Image Sensor |
Dimensions (L x W x H) | 24 x 33 x 17 cm (9.4 x 13 x 6.7 in.) |
Light Source | Xenon Flash |
Product Line | Microvolume |
Type | UV-Vis Spectrophotometer |
Voltage | 100/240 V |
Wavelength Range | 190 to 850 nm |
Measurement Time | 8 samples in ≤ 20 sec |
Pathlength | 1.0 mm, 0.2 mm, 0.1 mm (automatically adjusting) |
Power Consumption | 3 W at Idle; 15 W at Working Conditions |
Sample Volume (Metric) | 1 to 2 μL |
Spectral Resolution | ≤1.8 nm (FWHM at Hg 254 nm) |
Warranty | 1 Year |
Wavelength Accuracy | ± 1 nm |
Weight (English) | 8.2 lb. |
Weight (Metric) | 3.7 kg |
Unit Size | Each |
Display | Touchscreen |
Product Line | NanoDrop |
Type | Spectrophotometer with Fluorometer |
For Use With (Application) | Sample Concentration |
Storage Requirements | Room Temperature |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
NDEIGHT-PIP | Each | 21,800.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
NDEIGHT-E1PIP | Each | 24,300.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
NDEIGHT-LAPTOP | Each | 22,300.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
NDE-QBFLEX | Each | Request A Quote | - | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Thermo Scientific™ NanoDrop™ Eight Multichannel UV-Vis spectrophotometer measures 8 samples at a time in less than 20 seconds, enabling quick and efficient analysis of your sample quantity and quality. Built-in Thermo Scientific™ Acclaro™ Sample Intelligence technology can identify sample contaminants and report true sample concentrations. The Qubit Flex Fluorometer is a benchtop fluorometer designed for highly accurate quantification of DNA, RNA, microRNA, and protein. With the Qubit Flex Fluorometer you have the flexibility to directly measure fluorescence of up to eight samples simultaneously.
NanoDrop Eight spectrophotometer features
Acclaro Contaminant Identification ensures samples are high quality
Specialized software algorithms provide clear information about the true concentration of your nucleic acid or protein despite any possible interfering effects of sample contaminants, so you can understand your sample composition and ensure your downstream experiments go right the first time.
Qubit Flex Fluorometer features