NanoDrop™ Lite Plus Printer Accessory
NanoDrop™ Lite Plus Printer Accessory
Thermo Scientific™

NanoDrop™ Lite Plus Printer Accessory

Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.
Have Questions?
Catalog number NDLP-PRNTR-MOD
Price (USD)/ Each
1,248.00
-
Add to cart
Price (USD)/ Each
1,248.00
Add to cart
NanoDrop™ Lite Plus Printer Accessory
Catalog numberNDLP-PRNTR-MOD
Price (USD)/ Each
1,248.00
-
Add to cart

Use an optional printer with your NanoDrop™ Lite Plus Spectrophotometer. Print data on freezer-compatible labels.

Print sample data on a cryogenic label

  • Sample number
  • Date
  • Time of Measurement
  • Sample Name
  • Assay
  • Concentration
  • Purity Ratio

For use with the NanoDrop Lite Spectrophotometer

Specifications
DescriptionPrinter Accessory for NanoDrop Lite Plus
For Use With (Equipment)NanoDrop Lite Plus Spectrophotometer
Unit SizeEach