Invitrogen™
Neon™ NxT Electroporation System Pipette
Catalog number: NEON1P
Have Questions?
Invitrogen™
Neon™ NxT Electroporation System Pipette
Catalog number: NEON1P
Have Questions?
Catalog Number
NEON1P
Unit Size
1 pipette
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
NEON1P1 pipette
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Neon NxT Electroporation System Pipette is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1). Each system comes with a pipette. This pipette is offered separately for those needing an additional pipette or replacement for their existing pipette.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Tips
Neon™ NxT tips (Cat. No. N1025, N10025, N1096, N10096)
Product Line
Neon NxT
Type
Electroporation Pipette
Volume (Metric)
10, 100 μL
For Use With (Application)
Transfection
For Use With (Equipment)
Neon™ NxT Electroporation System