Invitrogen™
Neon™ NxT Electroporation System Pipette Station
Catalog number: NEON1PS
Have Questions?
Invitrogen™
Neon™ NxT Electroporation System Pipette Station
Catalog number: NEON1PS
Have Questions?
Catalog Number
NEON1PS
Unit Size
1 station
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
NEON1PS1 station
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Neon NxT Electroporation System Pipette Station is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1). Each system comes with a pipette station. This pipette station is offered separately for those needing an additional pipette station or replacement for their existing pipette station.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Compatible Tips
Neon™ NxT tips (Cat. No. N1025, N10025, N1096, N10096)
Type
Electroporation Pipette Station
For Use With (Application)
Transfection
For Use With (Equipment)
Neon™ NxT Electroporation System