Thermo Scientific™

Niton™ XL5 Plus Handheld XRF Analyzer

Catalog number: NITONXL5PLUS
Have Questions?
Thermo Scientific™

Niton™ XL5 Plus Handheld XRF Analyzer

Catalog number: NITONXL5PLUS
Have Questions?
Confidently perform elemental analysis with the Thermo Scientific™ Niton™ XL5 Plus handheld XRF analyzer; it delivers unprecedented performance even for lightweight elements. Identify pure metals and alloys, detect tramp elements, or obtain geochemical data. Industrial businesses rely on the Niton XL5 Plus analyzer for more than just low detection limits. It generates fast results and offers great versatility such as the ability to scan a broad range of materials with a single analyzer.
 
Catalog Number
NITONXL5PLUS
Unit Size
Each
Price (USD)
Full specifications
Data Storage512 MB internal system memory / 16 GB industrial grade storage; Stores approximately 130,000 readings with spectra (fewer if macro and micro images are saved)
Data TransferWiFi, USB-c
DescriptionNiton XL5 Plus Handheld XRF Analyzer
Detector TypeHigh count rate, high resolution, extra large area silicon drift detector (1μm graphene window). Detector ProGuard protection included.
Display TypeTilting, color, resistive touchscreen display
Optional Accessories
  • Portable test stand
  • Mini test stand
  • Backscatter shield
  • Hotwork stand off
  • Soil guard
  • Belt holster
  • Bluetooth printer
Power Consumption12V lithium-ion battery, or 12V DC, 3A, 3.6W power supply. Hot swap functionality keeps analyzer powered during battery replacements.
Security FeaturesPassword-protected user security
Standard Accessories
  • Locking shielded carrying case
  • Two (2) lithium-ion battery packs
  • One (1) 110/220 VAC battery charger/AC adaptor
  • Check samples
  • Safety lanyard
  • PC connection cable (USB)
System DetailsBuilt-in standardization and health check verify system integrity and operating conditions
Weight (English)2.8 lb. with battery
Weight (Metric)1.3 kg with battery
Data Entry
  • Touchscreen keyboard
  • User customizable data entry
  • Optional wireless remote barcode reader
LicensingVaries by region. Contact your local distributor.
Dimensions (L x W x H)9.54 x 8.19 x 2.67 in. (24.23 x 20.8 x 6.78 cm)
LanguagesEnglish, Chinese, Spanish, Portuguese, Russian, Japanese, German, Korean, French, Turkish, Italian
X-Ray Source Type
  • X-ray tube: Ag anode (6-50kV, 0-500uA, 5W max)
  • Filter: Six (6) position filter wheel for enhanced spectral range coverage
  • Current: Dynamically adjustable current for optimal sensitivity on every analysis
Spot Size
  • Standard: 8mm collimation
  • Optional: 3mm small-spot collimation
LibrariesDefault alloy libraries based on SAE, AISI, ASTM, AA, DIN, GB standards. Users may create, clone and edit libraries.
Operating Environment
  • Temperature: 0°C to 50°C (external fan recommended when ambient temperature is greater than 33°C)
  • Humidity: 10% to 90% relative humidity non-condensing
SoftwareSupport software: NitonConnect PC software
Global Positioning SystemInternal GPS and optional external GPS (via Bluetooth) GPS data included with sample information.
Operating SystemLinux
Certifications/ComplianceCE, RoHS, FCC, Industry Canada, Safety to IEC 61010-1:2010
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
NITONXL5PLUSFull specifications
EachRequest A Quote
Data Storage512 MB internal system memory / 16 GB industrial grade storage; Stores approximately 130,000 readings with spectra (fewer if macro and micro images are saved)
Data TransferWiFi, USB-c
DescriptionNiton XL5 Plus Handheld XRF Analyzer
Detector TypeHigh count rate, high resolution, extra large area silicon drift detector (1μm graphene window). Detector ProGuard protection included.
Display TypeTilting, color, resistive touchscreen display
Optional Accessories
  • Portable test stand
  • Mini test stand
  • Backscatter shield
  • Hotwork stand off
  • Soil guard
  • Belt holster
  • Bluetooth printer
Power Consumption12V lithium-ion battery, or 12V DC, 3A, 3.6W power supply. Hot swap functionality keeps analyzer powered during battery replacements.
Security FeaturesPassword-protected user security
Standard Accessories
  • Locking shielded carrying case
  • Two (2) lithium-ion battery packs
  • One (1) 110/220 VAC battery charger/AC adaptor
  • Check samples
  • Safety lanyard
  • PC connection cable (USB)
System DetailsBuilt-in standardization and health check verify system integrity and operating conditions
Weight (English)2.8 lb. with battery
Weight (Metric)1.3 kg with battery
Data Entry
  • Touchscreen keyboard
  • User customizable data entry
  • Optional wireless remote barcode reader
LicensingVaries by region. Contact your local distributor.
Dimensions (L x W x H)9.54 x 8.19 x 2.67 in. (24.23 x 20.8 x 6.78 cm)
LanguagesEnglish, Chinese, Spanish, Portuguese, Russian, Japanese, German, Korean, French, Turkish, Italian
X-Ray Source Type
  • X-ray tube: Ag anode (6-50kV, 0-500uA, 5W max)
  • Filter: Six (6) position filter wheel for enhanced spectral range coverage
  • Current: Dynamically adjustable current for optimal sensitivity on every analysis
Spot Size
  • Standard: 8mm collimation
  • Optional: 3mm small-spot collimation
LibrariesDefault alloy libraries based on SAE, AISI, ASTM, AA, DIN, GB standards. Users may create, clone and edit libraries.
Operating Environment
  • Temperature: 0°C to 50°C (external fan recommended when ambient temperature is greater than 33°C)
  • Humidity: 10% to 90% relative humidity non-condensing
SoftwareSupport software: NitonConnect PC software
Global Positioning SystemInternal GPS and optional external GPS (via Bluetooth) GPS data included with sample information.
Operating SystemLinux
Certifications/ComplianceCE, RoHS, FCC, Industry Canada, Safety to IEC 61010-1:2010
Unit SizeEach
Showing 1 of 1

Features

  • 5W X-ray tube
  • Proprietary large area drift detector for light element detection (Mg, Al, Si, P, S)
  • Lightweight design (2.8 lbs/ 1.3 kg)
  • Miniaturized nose
  • WiFi enabled
  • Macro and micro cameras
  • Hot swap battery
  • Tilting, color touchscreen
  • Directional keys
  • Password protected security
  • IP54 certified (splash/dust proof)

Applications

  • Verification of metals and alloys in manufacturing operations
  • Non-destructive field inspections for positive material identification
  • Point-and-shoot sorting at scrap recycling operations
  • Coating thickness measurements for single or binary metal layers
  • Precious metal assay of bullion and jewelry
  • Real-time geochemical analysis for mining exploration
  • On-site heavy metal screening of polluted soils
  • Screening of hazardous substances in consumer goods
  • Custom applications on demand

FAQ's

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text