NuPAGE™ MOPS SDS Running Buffer (20X)
NuPAGE™ MOPS SDS Running Buffer (20X)
Invitrogen™

NuPAGE™ MOPS SDS Running Buffer (20X)

NuPAGE MOPS SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Bis-Tris gels. It is recommended for separatingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
NP0001500 mL
NP0001025 L
Catalog number NP0001
Price (USD)
114.00
Each
Add to cart
Quantity:
500 mL
Price (USD)
114.00
Each
Add to cart
Ask our AI about this Product
NuPAGE MOPS SDS Running Buffer (20X) is formulated for running proteins on NuPAGE Bis-Tris gels. It is recommended for separating medium- to large-sized proteins.

Use the right buffer to optimize protein separations
NuPAGE MES SDS Running Buffer and NuPAGE MOPS SDS Running Buffer can both be used with NuPAGE Bis-Tris gels. MES has a lower pKa than MOPS, making the MES SDS Running Buffer faster than MOPS SDS running buffers. The difference in ion migration affects stacking and results in a difference in protein separation range between these buffers. Use of MOPS buffer allows proteins to run slower than when using MES buffer.

Compare protein migration patterns using MES and MOPs on NuPAGE Bis-Tris gels

See all available buffers and reagents available for SDS-PAGE

WARNING: Cancer – www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialRunning Buffer
Formulation50 mM MOPS, 50 mM Tris Base, 0.1% SDS, 1 mM EDTA, pH 7.7, 0.01-0.09% N,N-dimethylformamide
Recommended StorageContents: NuPAGE™ MOPS SDS Running Buffer (20X)
Storage: +4°C to 25°C
Concentration20X
Physical FormLiquid
Product LineNuPAGE
Quantity500 mL
pH7.7
Unit SizeEach
iBlot 3 Western Blot Transfer System

Introducing iBlot 3 Western Blot Transfer System

Featuring higher throughput and built-in cooling for consistent protein transfer
Learn more

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link