NuPAGE™ Transfer Buffer (20X)
NuPAGE™ Transfer Buffer (20X)
Invitrogen™

NuPAGE™ Transfer Buffer (20X)

NuPAGE Transfer Buffer (20X) is used to transfer proteins from NuPAGE Bis-Tris and NuPAGE Tris-Acetate gels to membranes for westernRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
NP0006125mL
NP000611L
Catalog number NP0006
Price (USD)
35.49
Each
In stock
Add to cart
Quantity:
125mL
Price (USD)
35.49
Each
Add to cart
Ask our AI about this Product
NuPAGE Transfer Buffer (20X) is used to transfer proteins from NuPAGE Bis-Tris and NuPAGE Tris-Acetate gels to membranes for western blotting. It maintains the neutral pH environment established during electrophoresis. The neutral pH protects against modification of amino acid side chains and is compatible with N-terminal protein sequencing using Edman degradation.

NuPAGE Antioxidant may be used with NuPAGE Transfer Buffer to enhance transfer of reduced proteins to membranes.

See all available buffers and reagents available for SDS-PAGE

For Research Use Only. Not for use in diagnostic procedures.
Specifications
BufferTransfer Buffers
Concentration20X
Gel CompatibilityNuPAGE™ Gels
Product LineNuPAGE™
Product TypeTransfer Buffer
Quantity125mL
Shelf Life6 Months
Shipping ConditionRoom Temperature
Gel TypeNuPAGE
Unit SizeEach
Contents & Storage
Store at room temperature. Guaranteed stable for 6 months unless otherwise specified in the product literature.
iBlot 3 Western Blot Transfer System

Introducing iBlot 3 Western Blot Transfer System

Featuring higher throughput and built-in cooling for consistent protein transfer
Learn more

Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Frequently asked questions (FAQs)

Can I prepare my protein sample with the reducing agent and store it for future use?

DTT is not stable, so it must be added and the reduction performed just prior to loading your samples.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

My LDS or SDS sample buffer precipitates when stored at 4 degrees C. Can I warm it up? Can I store it at room temperature?

Precipitation of the LDS or SDS at 4 degrees C is normal. Bring the buffer to room temperature and mix until the LDS/SDS goes into solution. If you do not want to wait for it to dissolve, you can store the sample buffer at room temperature.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

How are Bolt gels different than NuPAGE gels?

While they are both Bis-Tris based gels, the chemistries are very different since Bolt gels are optimized for western blotting. Another key difference is the wedge well design of the Bolt gels, which allows larger sample volumes to be loaded.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

What is the advantage of NuPAGE Gels over regular Tris-Glycine gels?

The neutral operating pH of the NuPAGE Gels and buffers provides following advantages over the Laemmli system:
-Longer shelf life of 8-12 months due to improved gel stability
-Improved protein stability during electrophoresis at neutral pH resulting in sharper band resolution and accurate results (Moos et al, 1998)
-Complete reduction of disulfides under mild heating conditions (70 degrees C for 10 min) and absence of cleavage of asp-pro bonds using the NuPAGE LDS Sample buffer (pH > 7.0 at 70 degrees C)
-Reduced state of the proteins maintained during electrophoresis and blotting of the proteins by the NuPAGE Antioxidant
Please refer to the following paper: Moos M Jr, Nguyen NY, Liu TY (1988) Reproducible High Yield Sequencing of Proteins Electrophoretically Separated and Transferred to an Inert Support. J Biol Chem 263:6005-6008.

Find additional tips, troubleshooting help, and resources within our Protein Gel 1D Electrophoresis Support Center.

How do I perform a semi-dry transfer of Tris-Glycine gels?

If using Invitrogen Semi-Dry Blotter (Cat. No. SD1000), follow instructions in the manual for it.

Here is the transfer protocol optimized for the Bio-Rad Semi-Dry Transfer Unit. NuPAGE transfer buffer can be used for Tris-Glycine gels.

1) Working transfer buffer: 10% methanol, 1:1,000 Antioxidant in 2X NuPAGE transfer buffer (Bis-Tris 50 mM and Bicine 50 mM). If you need to prepare 100 mL of the working buffer from the NuPAGE 20X Transfer Buffer (Cat. No. NP0006), mix the following: 10 mL of 20X transfer buffer, 10 mL of MeOH, 100 µL of Antioxidant, 80 mL of DI H2O.

2) Filter papers: The transfer buffer-soaked filter papers of the sandwich are the only reservoir in the Semi-Dry Transfer Cell. If Invitrogen pre-cut membrane/filter sandwiches are used, at least 2 extra filter papers (0.4 mm/filter in thickness) on each side of the gel (or membrane) are required. When assembling one gel/membrane sandwich, presoak 6 Invitrogen filter papers (or 2 thicker filter papers) and 1 membrane in working transfer buffer (prepared in step 1) and sandwich them on the top of the anode plate as follows: filter paper--filter paper--filter paper--membrane--gel--filter paper--filter paper--filter paper

3) Blotting conditions: We found 15 V for 15-30 min is optimal for NuPAGE transfer buffer in the Bio-Rad Semi-Dry Transfer Cell. Semi-dry transfer units from other manufacturers should be used according to unit's instructions.

4) For transfer of large proteins (100 kD or larger), pre-equilibrate the gel with 0.02-0.04% SDS in 2X transfer buffer without methanol for 10 min before assembling the sandwich. Please note that transferring Tris-Glycine gels using NuPAGE transfer buffer in the Bio-Rad Trans-Blot SD Semi-Dry Transfer Cell may be less efficient than using Tris-Glycine transfer buffer (Cat. No. LC3675) in the XCell II blot module (semi-wet).

Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
TFQAPO7QZCertificate of AnalysisJul 08, 2025NP0006
3187751Certificate of AnalysisJun 10, 2025NP00061
3178562Certificate of AnalysisMay 29, 2025NP00061
3136374Certificate of AnalysisApr 08, 2025NP00061
TESTPUBBSE/TSE CertificationMay 12, 2025NP0006
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (1)

Citations & References
Abstract
Matrix GLA protein, a regulatory protein for bone morphogenetic protein-2.
Authors: Zebboudj Amina F; Imura Minori; Boström Kristina;
Journal:J Biol Chem
PubMed ID:11741887
'Matrix GLA protein (MGP) has been identified as a calcification inhibitor in cartilage and vasculature. Part of this effect may be attributed to its influence on osteoinductive activity of bone morphogenetic protein-2 (BMP-2). To detect binding between MGP and BMP-2, we performed immunoprecipitation using MGP and BMP-2 tagged with FLAG ... More
1 total citations

Share catalog number, name or link

1x1 image pixel for data collection