You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Western Blot Products
General Western Blot Reagents
NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels)
Invitrogen™
NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels)
Catalog number:
NP0050
Related applications:
Protein Electrophoresis & Western Blotting
Have Questions?
Contact Us
Invitrogen™
NuPAGE™ MOPS SDS Buffer Kit (for Bis-Tris Gels)
Catalog number:
NP0050
Related applications:
Protein Electrophoresis & Western Blotting
Have Questions?
Contact Us
Catalog Number
NP0050
Unit Size
1 kit
Price (USD)
125.65
Online price
127.00
Availability
-
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Availability
Quantity
NP0050
1 kit
125.65
Online price
127.00
-
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The NuPAGE MOPS SDS Buffer Kit is designed for separation of medium- to large-size proteins on NuPAGE Bis-Tris gels and includes the following buffers:
• NuPAGE MOPS SDS Running Buffer (20X, 500 mL, Cat. No. NP0001)
• NuPAGE Sample Reducing Agent (10X, 250 μL, Cat. No. NP0004)
• NuPAGE Antioxidant (Cat. No. NP0005)
• NuPAGE LDS Sample Buffer (4X, 10 mL, Cat. No. NP0007)
Compare protein migration patterns using MES and MOPs on NuPAGE Bis-Tris gels
See all available buffers and reagents available for SDS-PAGE
WARNING:
Cancer –
www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel Type
SDS PAGE Gel
Shipping Condition
Room Temperature
Gel Compatibility
NuPAGE Bis-Tris Gel
Product Line
NuPAGE™
Product Type
Running Buffer
Quantity
1 kit
Contents & Storage
Contains
• NuPAGE™ MOPS SDS Running Buffer (20X, 500 ml)
• NuPAGE™ Sample Reducing Agent (10X, 250 μl) containing 500 mM Dithiothreitol
• NuPAGE™ Antioxidant (15 ml)
• NuPAGE™ LDS Sample Buffer (10 ml)
Store each at 4°C. Warm NuPAGE™ LDS Sample Buffer to 25°C before use.
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search