NuPAGE™ Bis-Tris IPG-well Welcome Pack
NuPAGE™ Bis-Tris IPG-well Welcome Pack
Invitrogen™
NuPAGE™ Bis-Tris IPG-well Welcome Pack
The NuPAGE Bis-Tris IPG-well Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need toRead more
Have Questions?
Catalog NumberQuantity
NP0330Z1 Welcome Pack
Catalog number NP0330Z
Price (TWD)
38,430.00
Online offer
Ends: 30-Jun-2025
43,800.00
Save 5,370.00 (12%)
Each
-
Add to cart
Quantity:
1 Welcome Pack
Price (TWD)
38,430.00
Online offer
Ends: 30-Jun-2025
43,800.00
Save 5,370.00 (12%)
Each
Add to cart
Ask our AI about this Product
The NuPAGE Bis-Tris IPG-well Welcome Pack provides all of the necessary NuPAGE gels, buffers, and reagents you will need to begin using the XCell SureLock Mini-Cell for your second-dimension separation in a 2D workflow. IPG-well gels are designed to easily accommodate 7-cm IPG strips and a molecular weight marker as well. The XCell SureLock Mini-Cell is a vertical mini-protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels.
The NuPAGE Bis-Tris Welcome Pack contains:
XCell SureLock Mini-Cell (Cat. No. EI0001)
NuPAGE Bis-Tris mini gels (2 boxes, 20 gels)
NuPAGE MES SDS Running Buffer, 20X (NP0002)
NuPAGE LDS Sample Buffer, 4X (NP0007)
NuPAGE Sample Reducing Agent, 10X (NP0009)
PageRuler Plus Prestained Protein Ladder, 10 to 250 kDa (26619)
Learn more about 2D electrophoresis ›
About the XCell SureLock Mini-Cell
The XCell SureLock Mini-Cell is a vertical mini protein gel electrophoresis system compatible with all Invitrogen mini precast gels and Surecast handcast gels. The Mini-Cell is simple, sturdy, and easy to use. Each XCell SureLock Mini-Cell can accommodate up to two gels per run. Run times may vary depending on buffer conditions and power supplies used.
ZOOM Bis-Tris IPG-Well Precast Gels
ZOOM gels are designed for optimal second dimension separation following isoelectric focusing. They feature an IPG well designed to easily accommodate 7-cm IPG strips and a molecular weight marker as well. Invitrogen IPG well gels are compatible with the XCell SureLock Mini Cell and the Mini Gel Tank.
About NuPAGE Bis-Tris gels
Invitrogen NuPAGE Bis-Tris Protein gels are precast polyacrylamide gels designed to give optimal separation of a wide range of proteins under denaturing conditions. Unlike traditional Tris-glycine gels, NuPAGE Bis-Tris gels have a neutral pH environment that minimizes protein modifications. Use NuPAGE Bis-Tris gels for preparing proteins for sequencing, mass spectrometry, and any other techniques where protein integrity is crucial. Also, use NuPAGE gels for optimal results during day-to-day use.
Specifications
For Use With (Application)Gel Electrophoresis, Isoelectric Focusing (IEF or IPG)
Gel Thickness1.0 mm
Product LineNuPAGE™
Quantity1 Welcome Pack
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Well DesignZoom IPG well
For Use With (Equipment)XCell SureLock Mini-Cell
Gel Percentage4 to 12%
Gel SizeMini
Gel TypeBis-Tris
Separation Range3.5 to 260 kDa
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection