Invitrogen™

Neon™ NxT Electroporation System Tubes

Catalog number: NT96
Have Questions?
Invitrogen™

Neon™ NxT Electroporation System Tubes

Catalog number: NT96
Have Questions?
Catalog Number
NT96
Unit Size
96 tubes
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
NT9696 tubes
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Neon NxT Electroporation System Tubes are designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON1). Each Neon NxT Electroporation System Kit for 25 x 2 reactions (Cat. Nos. N1025 and N10025) comes with eight electroporation tubes and each Neon NxT Electroporation System Kit for 96 x 2 reactions (Cat. Nos. N1096 and N10096) comes with 32 electroporation tubes. This pack of 96 tubes is offered for those wishing to use single tubes per reaction depending on their experimental setup.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format
Electroporation Pipette Tip
Transfection Technique
Electroporation
For Use With (Equipment)
Neon™ NxT Electroporation System
For Use With (Application)
Transfection

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text