Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200 assays (2 mL assay volume) |
Quantitation Range | 200 pg to 2 μg |
Product Line | OLIGREEN, Quant-iT |
Product Type | Quant-iT OliGreen ssDNA Kit |
Shipping Condition | Room Temperature |
Unit Size | 1 kit |
Detection Method | Fluorescence |
Excitation/Emission | 500/525 |
For Use With (Equipment) | Microplate Reader |
No. of Reactions | 200 assays (2 mL assay volume) |
Quantitation Range | 200 pg to 2 μg |
Product Line | OLIGREEN, Quant-iT |
Product Type | Quant-iT OliGreen ssDNA Reagent |
Shipping Condition | Room Temperature |
Unit Size | 1 mL |
Catalog Number | Specifications | Unit Size | Product Type | Quantitation Range | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
O11492 | 1 kit | Quant-iT OliGreen ssDNA Kit | 200 pg to 2 μg | 701.65 Online price 730.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
O7582 | 1 mL | Quant-iT OliGreen ssDNA Reagent | 200 pg to 2 μg | 656.65 Online price 678.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|