The green fluorescent Alexa Fluor™ 488 ovalbumin conjugate is a protein with a relatively low molecular weight (∼45,000 daltons) labeledRead more
Have Questions?
Catalog number O34781
Price (USD)/ 2 mg
155.65
Online exclusive
161.00
Save 5.35 (3%)
-
Add to cart
Price (USD)/ 2 mg
155.65
Online exclusive
161.00
Save 5.35 (3%)
Add to cart
Ovalbumin, Alexa Fluor™ 488 Conjugate
Catalog numberO34781
Price (USD)/ 2 mg
155.65
Online exclusive
161.00
Save 5.35 (3%)
-
Add to cart
The green fluorescent Alexa Fluor™ 488 ovalbumin conjugate is a protein with a relatively low molecular weight (∼45,000 daltons) labeled with the bright, photostable and pH-insensitive Alexa Fluor™ 488 dye.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity2 mg
ConjugateAlexa Fluor 488
Label TypeAlexa Fluor Dyes
Shipping ConditionRoom Temperature
FormLyophilized
Protein SubtypeAlbumin Proteins
Product LineAlexa Fluor™
Unit Size2 mg
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.