Xcalibur™ Software
Xcalibur™ Software
Actual product may vary
Thermo Scientific™
Xcalibur™ Software
Thermo Scientific™ Xcalibur™ software lets you analyze samples from a single interface, including instrument setup, method development, data acquisition, and more.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeIncludes
OPTON-30965Software4.3 + NIST Libraries
OPTON-30966Software4.3 Upgr fr 4.2/4.2 SP1
OPTON-30967Software4.3 Upgr fr 4.1 and older
Catalog number OPTON-30965
Price (USD)
-
Request A Quote
Type:
Software
Includes:
4.3 + NIST Libraries
Ask our AI about this Product
Control and process data from Thermo Scientific™ LC-MS systems and related instruments with easy-to-use Thermo Scientific™ Xcalibur™ software. Windows™-based Xcalibur software provides method setup, data acquisition, data processing, and data acquisition and processing integration to provide higher quality and more relevant data. Xcalibur software integrates seamlessly with application-specific Thermo Scientific software packages, and now includes both Thermo Scientific™ FreeStyle™ and Thermo Scientific™ Almanac™ web-based monitoring for cloud-enabled and cloud-independent functionality.
Efficient, simple data acquisition, interrogration and in-depth analysis
Xcalibur software provides efficient data acquisition, data processing, and results delivery, all via an easy-to-use interface.
  • Access your comprehensive toolset through the homepage, from included applications such as FreeStyle and Quan Browser to optional application-specific data processing tools to go seamlessly from method development to data acquisition, processing, and reporting
  • Ensure that you are always up-to-date with notifications when new version of software are available
  • Benefit from the web-based monitoring provided by cloud-based applications such as Almanac web-based monitoring to allow remote-monitoring of your systems and increase productivity
Specifications
For Use With (Equipment)Mass Spectrometers
Includes4.3 + NIST Libraries
Software4.3
TypeSoftware
Unit SizeEach
Dig into the complete capabilities of Xcalibur software
Dig into the complete capabilities of Xcalibur software
Find out more about Xcalibur software and its many included applications to streamline your laboratory workflows.
Learn more ›
Save 51% on various cell culture plasticware
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link