Thermo Scientific™
Mass Frontier™ Spectral Interpretation Software
Catalog number: OPTON-30978
Have Questions?
Thermo Scientific™
Mass Frontier™ Spectral Interpretation Software
Catalog number: OPTON-30978
Have Questions?
Transform your mass spectral data into knowledge quickly and easily using Thermo Scientific™ Mass Frontier™ spectral interpretation software. Take the challenge out of small molecule structural elucidation for metabolism, metabolomics, forensics, natural products, impurities, and degradants studies. High-quality, curated experimental spectral and fragmentation libraries simplify the deconvolution, evaluation, interpretation and management of infusion, and LC, nominal mass, or high resolution mass spectral data. Streamline building and curating your library and sharing it with your colleagues and collaborators. Mass Frontier software complements Thermo Scientific™ Compound Discoverer™ software for routine-based small molecule discovery and identification.
Catalog Number
OPTON-30978
Unit Size
Each
Description
Mass Frontier 8.0 SR1 w/Curator (one license)
Price (USD)
Full specifications
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 w/Curator (one license)
For Use With (Equipment)Mass Spectrometers
IncludesCurator and 1 License
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.0 upgrade for Compound Discoverer (one license).
Installer will check for valid previous Mass Frontier license.
Price (USD)
Full specifications
TypeSoftware Upgrade
Software8
DescriptionMass Frontier 8.0 upgrade for Compound Discoverer (one license).
Installer will check for valid previous Mass Frontier license.
For Use With (Equipment)Mass Spectrometers
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.0 SR1 (one license)
Price (USD)
Full specifications
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 (one license)
For Use With (Equipment)Mass Spectrometers
Includes1 License
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.0 SR1 (2+ licenses)
Price (USD)
Full specifications
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 (2+ licenses)
For Use With (Equipment)Mass Spectrometers
Includes2+ Licenses
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.0 SR1 w/Curator (2+ licenses)
Price (USD)
Full specifications
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 w/Curator (2+ licenses)
For Use With (Equipment)Mass Spectrometers
IncludesCurator and 2+ License
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.0 SR1 upgrade for Compound Discoverer (one license)
Installer will check for valid previous Mass Frontier license
Price (USD)
Full specifications
TypeSoftware Upgrade
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 upgrade for Compound Discoverer (one license)
Installer will check for valid previous Mass Frontier license
For Use With (Equipment)Mass Spectrometers
Includes1 License
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Curator License for MF 8.1 (single)
Price (USD)
Full specifications
TypeSoftware
DescriptionCurator License for MF 8.1 (single)
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Curator License for MF 8.1 (multiple)
Price (USD)
Full specifications
TypeSoftware
DescriptionCurator License for MF 8.1 (multiple)
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.1
Price (USD)
Full specifications
TypeSoftware
DescriptionMass Frontier 8.1
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mass Frontier 8.1 with Curator license
Price (USD)
Full specifications
TypeSoftware
DescriptionMass Frontier 8.1 with Curator license
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
OPTON-30978Full specifications
EachMass Frontier 8.0 SR1 w/Curator (one license)Request A Quote
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 w/Curator (one license)
For Use With (Equipment)Mass Spectrometers
IncludesCurator and 1 License
Unit SizeEach
OPTON-30920Full specifications
EachMass Frontier 8.0 upgrade for Compound Discoverer (one license).
Installer will check for valid previous Mass Frontier license.
Request A Quote
TypeSoftware Upgrade
Software8
DescriptionMass Frontier 8.0 upgrade for Compound Discoverer (one license).
Installer will check for valid previous Mass Frontier license.
For Use With (Equipment)Mass Spectrometers
Unit SizeEach
OPTON-30976Full specifications
EachMass Frontier 8.0 SR1 (one license)Request A Quote
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 (one license)
For Use With (Equipment)Mass Spectrometers
Includes1 License
Unit SizeEach
OPTON-30977Full specifications
EachMass Frontier 8.0 SR1 (2+ licenses)Request A Quote
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 (2+ licenses)
For Use With (Equipment)Mass Spectrometers
Includes2+ Licenses
Unit SizeEach
OPTON-30979Full specifications
EachMass Frontier 8.0 SR1 w/Curator (2+ licenses)Request A Quote
TypeSoftware
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 w/Curator (2+ licenses)
For Use With (Equipment)Mass Spectrometers
IncludesCurator and 2+ License
Unit SizeEach
OPTON-30984Full specifications
EachMass Frontier 8.0 SR1 upgrade for Compound Discoverer (one license)
Installer will check for valid previous Mass Frontier license
Request A Quote
TypeSoftware Upgrade
Software8.0 SR1
DescriptionMass Frontier 8.0 SR1 upgrade for Compound Discoverer (one license)
Installer will check for valid previous Mass Frontier license
For Use With (Equipment)Mass Spectrometers
Includes1 License
Unit SizeEach
B51001149Full specifications
EachCurator License for MF 8.1 (single)Request A Quote
TypeSoftware
DescriptionCurator License for MF 8.1 (single)
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
B51001150Full specifications
EachCurator License for MF 8.1 (multiple)Request A Quote
TypeSoftware
DescriptionCurator License for MF 8.1 (multiple)
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
B51000940Full specifications
EachMass Frontier 8.1Request A Quote
TypeSoftware
DescriptionMass Frontier 8.1
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
B51000941Full specifications
EachMass Frontier 8.1 with Curator licenseRequest A Quote
TypeSoftware
DescriptionMass Frontier 8.1 with Curator license
For Use With (Equipment)Mass Spectrometer
Unit SizeEach
Showing 10 of 12Show more
See what's really there
  • Deconvolute your nominal mass or high resolution LC-MSn or infusion data.
  • Capture all of your data, no matter how complex, with simple interrogation of all components.
Know more unknowns
  • Transform your small molecule unknown data into actionable knowledge.
  • Search mzCloud™ mass spectral library, the world's most comprehensive and structurally diverse fully curated mass spectral database.
  • Maximize the utility of your MSn data for the confident identification of unknown with spectral tree searching.
  • Enhance your confidence in proposed unknown candidate structures by using Thermo Scientific™ mzLogic™ algorithm, based on similarity searching to millions of fragment annotated spectra.
Go from spectra to structure
  • The HighChem Fragmentation Library™ includes >54,000 fragmentation schemes, ∼ 220,000 individual reactions, and 256,000 chemical structures, all from peer-reviewed literature.
  • Spectral trees are fully annotated with fragmentation structures.
  • FISh (fragment ion search) detection enables detection of structurally related compounds, indicating the site of transformations for drug metabolites.
Build your pathways
  • Interactively search more than 370 curated biochemical pathways with Thermo Scientific™ Metabolika™ pathway mapping.
  • Create and understand fragmentation pathways and mechanisms using advanced fragmentation scheme knowledge base drawn from more than 99% of published fragmentation pathways.
Store, share and utilize your knowledge
  • Translate your data into actionable knowledge: create, curate and store your own spectral libraries and pathways.
  • Ensure the highest quality proprietary libraries with full curation and mass recalibration tools.
Specifications, terms, and pricing are subject to change. Not all products are available in all countries. Please consult your local sales representative for details.