Thermo Scientific™

BioPharma Finder™ Software for the Characterization of Biotherapeutics

Catalog number: OPTON-30988
Have Questions?
Thermo Scientific™

BioPharma Finder™ Software for the Characterization of Biotherapeutics

Catalog number: OPTON-30988
Have Questions?
Simplify comprehensive biotherapeutic attribute characterization with advanced capabilities to increase throughput and confidence for BioPharma workflows through built for purpose data processing, curation, and reporting in a connected environment to streamline and enable collaboration on a local and global scale with Thermo Scientific™ BioPharma Finder™ software. Take full advantage of the high-quality, HRAM data produced by Thermo Scientific™ Orbitrap™ mass spectrometers to screen, identify, and characterize intact proteins and oligonucleotides with higher productivity and confidence.
 
Catalog Number
OPTON-30988
Unit Size
Each
Description
BioPharma Finder 5.2 Oligonucleotide Workflows (Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Oligonucleotide Workflows (Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 All Workflows (Peptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 All Workflows (Peptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Peptide Mapping Only
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Peptide Mapping Only
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Intact Mass and Top Down Only
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Intact Mass and Top Down Only
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass and Top Down
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Intact
(Upgrade from Intact Mass and Top Down Only – Adds Oligonucleotide Analysis and Oligonucleotide Calculator)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Intact
(Upgrade from Intact Mass and Top Down Only – Adds Oligonucleotide Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculators
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Peptide Mapping
(Upgrade from Peptide Mapping – Adds Oligonucleotide Analysis, Intact Mass, Top Down, and Oligonucleotide Calculator)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Peptide Mapping
(Upgrade from Peptide Mapping – Adds Oligonucleotide Analysis, Intact Mass, Top Down, and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Add on Peptide Mapping to Existing Protein Workflows
(Upgrade from Intact Mass and Top Down Only – Adds Peptide Mapping Workflows)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Peptide Mapping to Existing Protein Workflows
(Upgrade from Intact Mass and Top Down Only – Adds Peptide Mapping Workflows)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass and Top Down
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Add on Intact and Top Down to Existing Peptide Mapping
(Upgrade from Peptide Mapping Only – Adds Intact Mass and Top Down Workflows)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Intact and Top Down to Existing Peptide Mapping
(Upgrade from Peptide Mapping Only – Adds Intact Mass and Top Down Workflows)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass and Top Down
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade 4.X All workflows
(Upgrades from 4.0 or 4.1 Peptide Mapping, Intact Mass, Top Down, and Oligonucleotide Analysis to current version and adds Oligonucleotide Calculator)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade 4.X All workflows
(Upgrades from 4.0 or 4.1 Peptide Mapping, Intact Mass, Top Down, and Oligonucleotide Analysis to current version and adds Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
BioPharma Finder 5.2 Upgrade 3.X All Protein Analysis Workflows
(Upgrade from 3.X Peptide Mapping, Intact Mass, and Top Down)
Price (USD)
Full specifications
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade 3.X All Protein Analysis Workflows
(Upgrade from 3.X Peptide Mapping, Intact Mass, and Top Down)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass and Top Down
Unit SizeEach
Showing 10 of 36Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
OPTON-30988Full specifications
EachBioPharma Finder 5.2 Oligonucleotide Workflows (Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Oligonucleotide Workflows (Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top-Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
OPTON-30985Full specifications
EachBioPharma Finder 5.2 All Workflows (Peptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 All Workflows (Peptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
OPTON-30986Full specifications
EachBioPharma Finder 5.2 Peptide Mapping OnlyRequest A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Peptide Mapping Only
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping
Unit SizeEach
OPTON-30987Full specifications
EachBioPharma Finder 5.2 Intact Mass and Top Down OnlyRequest A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Intact Mass and Top Down Only
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass and Top Down
Unit SizeEach
OPTON-30989Full specifications
EachBioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Intact
(Upgrade from Intact Mass and Top Down Only – Adds Oligonucleotide Analysis and Oligonucleotide Calculator)
Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Intact
(Upgrade from Intact Mass and Top Down Only – Adds Oligonucleotide Analysis and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesIntact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculators
Unit SizeEach
OPTON-30990Full specifications
EachBioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Peptide Mapping
(Upgrade from Peptide Mapping – Adds Oligonucleotide Analysis, Intact Mass, Top Down, and Oligonucleotide Calculator)
Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Oligonucleotide Workflows to Existing Peptide Mapping
(Upgrade from Peptide Mapping – Adds Oligonucleotide Analysis, Intact Mass, Top Down, and Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
OPTON-30991Full specifications
EachBioPharma Finder 5.2 Add on Peptide Mapping to Existing Protein Workflows
(Upgrade from Intact Mass and Top Down Only – Adds Peptide Mapping Workflows)
Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Peptide Mapping to Existing Protein Workflows
(Upgrade from Intact Mass and Top Down Only – Adds Peptide Mapping Workflows)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass and Top Down
Unit SizeEach
OPTON-30992Full specifications
EachBioPharma Finder 5.2 Add on Intact and Top Down to Existing Peptide Mapping
(Upgrade from Peptide Mapping Only – Adds Intact Mass and Top Down Workflows)
Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Add on Intact and Top Down to Existing Peptide Mapping
(Upgrade from Peptide Mapping Only – Adds Intact Mass and Top Down Workflows)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass and Top Down
Unit SizeEach
OPTON-30993Full specifications
EachBioPharma Finder 5.2 Upgrade 4.X All workflows
(Upgrades from 4.0 or 4.1 Peptide Mapping, Intact Mass, Top Down, and Oligonucleotide Analysis to current version and adds Oligonucleotide Calculator)
Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade 4.X All workflows
(Upgrades from 4.0 or 4.1 Peptide Mapping, Intact Mass, Top Down, and Oligonucleotide Analysis to current version and adds Oligonucleotide Calculator)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass, Top Down, Oligonucleotide Analysis, and Oligonucleotide Calculator
Unit SizeEach
OPTON-30994Full specifications
EachBioPharma Finder 5.2 Upgrade 3.X All Protein Analysis Workflows
(Upgrade from 3.X Peptide Mapping, Intact Mass, and Top Down)
Request A Quote
TypeSoftware
Software5.2
DescriptionBioPharma Finder 5.2 Upgrade 3.X All Protein Analysis Workflows
(Upgrade from 3.X Peptide Mapping, Intact Mass, and Top Down)
For Use With (Equipment)Mass Spectrometers
IncludesPeptide Mapping, Intact Mass and Top Down
Unit SizeEach
Showing 10 of 36Show more

Workflow-driven experiment creation, method processing, and result review
Intact protein workflow:

  • Rapidly measure the intact molecular weight of biotherapeutics, including proteins and oligonucleotides, for structural confirmation and characterization
  • Confidently determine deconvoluted molecular weights
  • Target protein sequence matching to identify n-linked glycosylations and other common modifications
  • Identify Antibody Drug Conjugates (ADC) using sliding window algorithm
  • Batch-to-batch comparison

Peptide mapping workflow:

  • Confirm amino acid sequence with a novel MS2 prediction algorithm, providing extra confidence for sequence assignments
  • Identify the site and type of expected and unknown PTMs
  • Map disulfide bonds
  • Detect low-level impurities and sequence variants
  • Perform error-tolerant searches for unexpected modifications
  • Create de novo sequence searches

Multi Attribute Methods (MAM):

  • MS-based MAM for characterization and relative quantification of PTMs
  • Features streamlined processes ranging from discovery peptide mapping to critical quality attributes (CQAs) monitoring in GMP environments

Top down workflow:

  • Features a simple workflow for sequencing intact protein molecules using ProSightBP as the core algorithm
  • Includes multiple raw file comparison with combined interactive fragmentation coverage maps
  • Supports multiple modes of fragmentation CID, HCD, ETD, EThcD and UVPD

Oligonucleotide workflow:

  • Support for oligonucleotide sequences with built in customized building block subunits, including base, backbone linker, 2' ribose, 3' terminal and 5' terminal
  • Automated annotation and confirmation of oligonucleotide sequence with a novel MS2 prediction algorithm, providing extra confidence in oligonucleotide sequencing

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text