Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Simplify comprehensive biotherapeutic attribute characterization with advanced capabilities to increase throughput and confidence in a connected environment.
Catalog Number | Includes |
---|---|
OPTON-30999 | Oligonucleotide, Intact Mass, Top-Down Analysis, and Oligonucleotide Calculator |
OPTON-30985 | Peptide Mapping, Oligonucleotide, Intact Mass, Top-Down Analysis, and Oligonucleotide Calculator |
OPTON-31029 | Peptide Mapping, Intact Mass, and Top-Down Analysis |
OPTON-30988 | Oligonucleotide, Intact Mass, Top-Down Analysis, and Oligonucleotide Calculator |
OPTON-30986 | Peptide Mapping |
OPTON-30987 | Intact Mass and Top Down Analysis |
B51000981 | 5.3 Oligo Calculator |
OPTON-31069 | Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator |
OPTON-31068 | Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator |
OPTON-30993 | Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator |
B51001849 | 5.3 Upgrade 5.X Prot Workflow |
OPTON-30989 | Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator |
OPTON-30990 | Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator |
OPTON-30991 | Peptide Mapping, Intact Mass, and Top Down Analysis |
OPTON-30992 | Peptide Mapping, Intact Mass, and Top Down Analysis |
B51001858 | 5.3 Upgrade 3.X All Workflows |
OPTON-30995 | Peptide Mapping, Oligonucleotide, Intact Mass, Top Down Analysis, and Oligonucleotide Calculator |
OPTON-31095 | Peptide Mapping, Intact Mass, and Top Down Analysis |
OPTON-30994 | Peptide Mapping, Intact Mass, and Top Down Analysis |
OPTON-31030 | Peptide Mapping, Intact Mass, and Top Down Analysis |
B51001850 | 5.3 Upgrd 5.X Oligo Workflow |
B51001855 | 5.3 Upgrd 4.X Oligo Workflow |
B51001859 | 5.3 Add Oligo WF from 3.X |
B51001860 | 5.3 Add Oligo WF from 2.0 |
B51001854 | 5.3 Upgrade 5.X Pep Mapping |
B51001857 | 5.3 Upgrade 4.X Pep Mapping |
OPTON-31033 | Peptide Mapping |
OPTON-31034 | Peptide Mapping |
B51001852 | 5.3 Upgrade 5.X Intact |
B51001856 | 5.3 Upgrade 4.X Intact |
OPTON-31031 | Intact Mass And Top Down Analysis |
OPTON-31032 | Intact Mass And Top Down Analysis |
OPTON-30996 | Peptide Mapping, Oligonucleotide, Intact Mass, Top-Down Analysis, and Oligonucleotide Calculator |
OPTON-30997 | Peptide Mapping |
OPTON-30998 | Intact Mass and Top Down Analysis |
OPTON-31035 | Peptide Mapping, Intact Mass, and Top-Down Analysis |
Thermo Scientific™ BioPharma Finder™ software offers purpose-built data processing, curation, and reporting in a connected environment, enabling streamlined collaboration on a local and global scale. Simplify the characterization of biotherapeutic attributes with advanced capabilities that increase efficiency and confidence in BioPharma workflows. Leverage the high quality HRAM data generated by Thermo Scientific™ Orbitrap™ mass spectrometers, to efficiently screen, identify, and characterize proteins and oligonucleotides with greater productivity and confidence. Experience the power of BioPharma Finder software connected to the Thermo Scientific™ Ardia™ Platform to unlock enhanced features including data security, remote accessibility, collaboration tools, and interactive reporting, all tailored to streamline your BioPharma workflows.
Workflow-driven characterization, method processing, and result review
Peptide mapping workflow:
Oligonucleotide workflow:
Intact protein workflow:
Top down workflow:
Ardia Platform:
BioPharma Finder software streamlines confident characterization of protein and oligonucleotide based therapeutics
Share catalog number, name or link