Thermo Scientific™
Compound Discoverer™ Software
Catalog number: OPTON-31063
Have Questions?
Thermo Scientific™
Compound Discoverer™ Software
Catalog number: OPTON-31063
Have Questions?
Streamline and customize high-resolution accurate-mass data analysis for small molecule workflows with Thermo Scientific™ Compound Discoverer™ software. Compound Discoverer software simplifies and reduces processing, enabling you to go from data to insights faster. Leverage easy to use pre-built workflow templates or fully customizable node-based workflows, integrated compound identification capabilities, and statistical analysis tools for streamlined data analyses to deliver confident results to drive your decision making.
Catalog Number
OPTON-31063
Unit Size
Each
Description
Bundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (2+ licenses)
Includes
Compound Discoverer 3.3 software and Mass Frontier 8.0 with Curator
Price (USD)
Full specifications
TypeSoftware Bundle
Software3.3
DescriptionBundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (2+ licenses)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software and Mass Frontier 8.0 with Curator
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Compound Discoverer 3.3 – Full Version (one license)
Includes
Compound Discoverer 3.3 software
Price (USD)
Full specifications
TypeSoftware
Software3.3
DescriptionCompound Discoverer 3.3 – Full Version (one license)
IncludesCompound Discoverer 3.3 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Compound Discoverer 3.3 – Full Version (2 or more licenses)
Includes
Compound Discoverer 3.3 software
Price (USD)
Full specifications
TypeSoftware
Software3.3
DescriptionCompound Discoverer 3.3 – Full Version (2 or more licenses)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Compound Discoverer 3.3 to Upgrade from Compound Discoverer Version 1.0/2.0/2.1. The installer will check for a valid previous CD 1.0/2.0/2.1 license.
Includes
Compound Discoverer 3.3 software
Price (USD)
Full specifications
TypeSoftware Upgrade
Software3.3
DescriptionCompound Discoverer 3.3 to Upgrade from Compound Discoverer Version 1.0/2.0/2.1. The installer will check for a valid previous CD 1.0/2.0/2.1 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Compound Discoverer 3.3 to Upgrade from Compound Discoverer Version 3.0/3.1/3.2. The installer will check for a valid previous CD 3.0/3.1/3.2 license.
Includes
Compound Discoverer 3.3 software
Price (USD)
Full specifications
TypeSoftware Upgrade
Software3.3
DescriptionCompound Discoverer 3.3 to Upgrade from Compound Discoverer Version 3.0/3.1/3.2. The installer will check for a valid previous CD 3.0/3.1/3.2 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared
IncludesCompound Discoverer 3.3 software
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Bundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (one license)
Includes
Compound Discoverer 3.3 software and Mass Frontier 8.0 with Curator
Price (USD)
Full specifications
TypeSoftware Bundle
Software3.3
DescriptionBundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (one license)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software and Mass Frontier 8.0 with Curator
Unit SizeEach
Showing 6 of 6
Catalog NumberSpecificationsUnit SizeDescriptionIncludesPrice (USD)
OPTON-31063Full specifications
EachBundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (2+ licenses)Compound Discoverer 3.3 software and Mass Frontier 8.0 with CuratorRequest A Quote
TypeSoftware Bundle
Software3.3
DescriptionBundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (2+ licenses)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software and Mass Frontier 8.0 with Curator
Unit SizeEach
OPTON-31055Full specifications
EachCompound Discoverer 3.3 – Full Version (one license)Compound Discoverer 3.3 softwareRequest A Quote
TypeSoftware
Software3.3
DescriptionCompound Discoverer 3.3 – Full Version (one license)
IncludesCompound Discoverer 3.3 software
Unit SizeEach
OPTON-31056Full specifications
EachCompound Discoverer 3.3 – Full Version (2 or more licenses)Compound Discoverer 3.3 softwareRequest A Quote
TypeSoftware
Software3.3
DescriptionCompound Discoverer 3.3 – Full Version (2 or more licenses)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software
Unit SizeEach
OPTON-31060Full specifications
EachCompound Discoverer 3.3 to Upgrade from Compound Discoverer Version 1.0/2.0/2.1. The installer will check for a valid previous CD 1.0/2.0/2.1 license.Compound Discoverer 3.3 softwareRequest A Quote
TypeSoftware Upgrade
Software3.3
DescriptionCompound Discoverer 3.3 to Upgrade from Compound Discoverer Version 1.0/2.0/2.1. The installer will check for a valid previous CD 1.0/2.0/2.1 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software
Unit SizeEach
OPTON-31061Full specifications
EachCompound Discoverer 3.3 to Upgrade from Compound Discoverer Version 3.0/3.1/3.2. The installer will check for a valid previous CD 3.0/3.1/3.2 license.Compound Discoverer 3.3 softwareRequest A Quote
TypeSoftware Upgrade
Software3.3
DescriptionCompound Discoverer 3.3 to Upgrade from Compound Discoverer Version 3.0/3.1/3.2. The installer will check for a valid previous CD 3.0/3.1/3.2 license.
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared
IncludesCompound Discoverer 3.3 software
Unit SizeEach
OPTON-31062Full specifications
EachBundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (one license)Compound Discoverer 3.3 software and Mass Frontier 8.0 with CuratorRequest A Quote
TypeSoftware Bundle
Software3.3
DescriptionBundle: Compound Discoverer 3.3 and Mass Frontier 8.0 SR1 with Curator (one license)
For Use With (Application)ICP-OES, ICP-MS, Noble Gas MS, and Isotope Ratio Infrared Spectrometry
IncludesCompound Discoverer 3.3 software and Mass Frontier 8.0 with Curator
Unit SizeEach
Showing 6 of 6
Know more unknowns
  • Identify compounds using automatic multiple database and spectral library search tools including mzCloud™, Chemspider™, KEGG, and BioCyc, and local database search tools such as the mzVault™ spectral library or mass lists.
  • Access the latest "auto-processed" mzCloud libraries and utilize both DIA and DDA scans for searching.
  • Determine elemental composition using high resolution fine isotopes, MS/MS, and MSn data.
  • Enables data analysis of high resolution, accurate mass Orbitrap mass spectrometers coupled to either liquid chromatography (LC) or gas chromatography (GC) instrumentation.
Workflow flexibility to match analytical requirements
Compound Discoverer software offers complete flexibility, making it easy to assemble a workflow that matches the analytical requirements of your analysis.
  • Workflows are modular and assembled from a suite of advanced algorithms (nodes).
  • Customizable workflow templates for different applications are included with the software for ease-of-use.
  • Workflows can be enhanced with custom nodes to meet the needs of your most challenging research.
Advanced data processing and visualization
  • Increase certainty using multi-factorial Peak Quality factors to reveal, identify, and quantify more compounds with confidence.
  • Biological pathway analysis with high quality data processing and statistical analysis—support for stable isotope labeling (“flux analysis”) workflows to analyze differences in samples.
  • Find real differences between sample sets with heat map-like tables, interactive PCA, partial least squares discriminant analysis (PLS-DA), etc.
  • Visualization of spectral trees along with fragmentation annotations and mzLogic algorithm for ranking chemical database hits using spectral library similarities for structural elucidation.
  • Molecular networks to explore relationships between compounds.