Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Perform comprehensive analysis of top-down proteomics data with Thermo Scientific™ ProSightPD™ software. ProSightPD features robust proteomic search engines for proteoform characterization, including proteoforms with PTMs or truncations. ProSightPD software accommodates proteomic data generated by fragmentation techniques present in any Thermo Scientific mass spectrometer. ProSightPD software is the only proteomics software that allows the user to easily search MS/MS data against proteome databases containing the complete biological complexity found in UniProt.
Upgrade ProSightPD before 4.1 to ProSightPD 4.2
Type | Software |
Software | 4.2 |
Description | Upgrade ProSightPD before 4.1 to ProSightPD 4.2 |
For Use With (Equipment) | Mass Spectrometers |
Includes | p>Thermo Scientific ProSight PD 4.2 software |
Unit Size | Each |
Type | Software |
Software | 2 |
Description | Thermo Scientific ProSightPD software. |
For Use With (Equipment) | Mass Spectrometers |
Unit Size | Each |
Type | Software |
Software | 2 |
Description | ProSightPD software. |
For Use With (Equipment) | Mass Spectrometers |
Unit Size | Each |
Type | Software |
Software | 4.2 |
Description | ProSightPD 4.2 software |
For Use With (Equipment) | Mass Spectrometers |
Includes | Thermo Scientific ProSight PD 4.2 software |
Unit Size | Each |
Upgrade ProSightPD 4.1 to ProSightPD 4.2
Type | Software |
Software | 4.2 |
Description | Upgrade ProSightPD 4.1 to ProSightPD 4.2 |
For Use With (Equipment) | Mass Spectrometers |
Includes | Thermo Scientific ProSight PD 4.2 software |
Unit Size | Each |
Upgrade ProSightPD 4.2 from ProSightPC
Type | Software |
Software | 4.2 |
Description | Upgrade ProSightPD 4.2 from ProSightPC |
For Use With (Equipment) | Mass Spectrometers |
Includes | Thermo Scientific ProSight PD 4.2 software |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
OPTON-31092 | Each | Upgrade ProSightPD before 4.1 to ProSightPD 4.2 | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
OPTON-30903 | Each | Thermo Scientific ProSightPD software. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
OPTON-30918 | Each | ProSightPD software. | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
OPTON-31090 | Each | ProSightPD 4.2 software | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
OPTON-31091 | Each | Upgrade ProSightPD 4.1 to ProSightPD 4.2 | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
OPTON-31093 | Each | Upgrade ProSightPD 4.2 from ProSightPC | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|