PEEK™ Tee High-Pressure HPLC Connection, 0.020 in. through hole, <0.57µL swept volume
PEEK&trade; Tee High-Pressure HPLC Connection, 0.020 in. through hole, &#60;0.57&micro;L swept volume
Thermo Scientific™
PEEK™ Tee High-Pressure HPLC Connection, 0.020 in. through hole, <0.57µL swept volume
Ensure reliable high-pressure connections with biocompatible Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections.
Have Questions?
Catalog number P-727
Price (USD)
85.00
Each
Add to cart
Price (USD)
85.00
Each
Add to cart
Minimize system dead volumes and ensure reliable high-pressure connections in your HPLC system with Thermo Scientific™ PEEK™ Unions, Tees and Crosses for High-Pressure HPLC Connections. The material is biocompatible for a wide range of applications.
  • Absolute zero or low dead volume formats
  • Biocompatible
Specifications
TypeTee
Diameter (English) Inner0.02 in.
DescriptionPEEK Tee;<0.57 uL Swept Volume
Diameter (Metric) Inner12.45 mm
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link