Invitrogen™

PowerLoad™ Concentrate, 100X

Catalog number: P10020
Have Questions?
Invitrogen™

PowerLoad™ Concentrate, 100X

Catalog number: P10020
Have Questions?
Catalog Number
P10020
Unit Size
5 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P100205 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
PowerLoad™ is an optimized formulation of nonionic, Pluronic™ surfactant polyols for the solubilization of water-insoluble dyes and other materials in physiological media. These surfactants, for instance Pluronic™ F-127, have been used to help disperse acetoxymethyl (AM) esters of fluorescent ion indicators such as fluo-4, fura-2, indo-1, fluo-3, and SBFI; they appear to be required for loading of other dyes (e.g. SBFI-AM or PBFI-AM). The use of PowerLoad™ is optional with red shifted calcium indicators and other large molecular weight AM ester dyes, and may also be useful for dispersing other lipophilic probes. The concentration of Pluronic™ surfactants in PowerLoad™ is less than 0.2%. PowerLoad™ is effective in combination with water soluble Probenecid (P36400) to aid AM ester dye-loading and retention in cells that actively extrude the de-acetylated form through anion pumps. Together, these reagents allow for maximal loading of dyes with a minimum of effort in both imaging and high throughput screening (HTS) applications. Appropriate controls should be performed to make certain that PowerLoad™ is not altering the membrane properties of the cell.

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Recommended Storage
Store in refrigerator (2–8°C).
Quantity
5 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text