Invitrogen™

Protein A, Alexa Fluor™ 488 conjugate

Catalog number: P11047
Have Questions?
Invitrogen™

Protein A, Alexa Fluor™ 488 conjugate

Catalog number: P11047
Have Questions?
Catalog Number
P11047
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P110471 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Our Alexa Fluor 488 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, green-fluorescent probe (absorption/emission maxima ∼495/519 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
Alexa Fluor 488
Quantity
1 mg
Product Line
Alexa Fluor™
Form
Lyophilized Powder
Protein Subtype
Protein A
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text