Invitrogen™

Protein G, Alexa Fluor™ 488 conjugate

Catalog number: P11065
Have Questions?
Invitrogen™

Protein G, Alexa Fluor™ 488 conjugate

Catalog number: P11065
Have Questions?
Catalog Number
P11065
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P110651 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Alexa Fluor 488 protein G conjugate exhibits bright, green fluorescence (absorption/emission ∼495/519 nm). Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Conjugate
Alexa Fluor 488
Quantity
1 mg
Product Line
Alexa Fluor™
Form
Lyophilized Powder
Shipping Condition
Room Temperature

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text