Invitrogen™

PBFI, Tetraammonium Salt, cell impermeant

Catalog number: P1265MP
Have Questions?
Invitrogen™

PBFI, Tetraammonium Salt, cell impermeant

Catalog number: P1265MP
Have Questions?
Catalog Number
P1265MP
also known as P-1265MP
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P1265MP
also known as P-1265MP
1 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
PBFI is a potassium-sensitive molecule used measure intracellular K+ fluxes in animal cells and in plant cells and vacuoles. Although the selectivity of PBFI for K+ is less than that of calcium indicators such as fura-2, it is sufficient for the detection of physiological concentrations of K+ in the presence of other monovalent cations. The spectral response of PBFI upon ion binding permit excitation ratio measurements, and this indicator can be used with the same optical filters and equipment used for fura-2.

Learn more about ion indicators including calcium, potassium, pH, and membrane potential indicators ›

Fluorescent Ion Indicators Specifications:
• Label (Ex/Em): PBFI (∼340,380/500 nm)
• Lyophilized product may be dissolved in distilled water or aqueous buffer for use
• Product is typically loaded into cells by diffusion from a patch pipette for correlated fluorescence imaging and electrophysiological recording


Selectivity Considerations and Cell Loading Strategies
The Kd of PBFI for K+ is strongly dependent on whether Na+ is present, with a value of 5.1 mM in the absence of Na+ and 44 mM in solutions with a combined Na+ and K+ concentration of 135 mM (which approximates physiological ionic strength). In buffers in which the Na+ is replaced by tetramethylammonium chloride, the Kd of PBFI for K+ is 11 mM; choline chloride and N-methylglucamine are two other possible replacements for Na+ in the medium. Although PBFI is only 1.5-fold more selective for K+ than for Na+, this selectivity is often sufficient because intracellular K+ concentrations are normally about 10 times higher than Na+ concentrations.

PBFI is available as both cell-impermeant acid salt (P1265MP) and as cell-permeant acetoxymethyl (AM) esters (P1267MP). The anionic acid forms can be loaded into cells using our Influx™ pinocytic cell-loading reagent (I14402, Chelators, Calibration Buffers, Ionophores and Cell-Loading Reagents—Section 19.8), or by microinjection, patch-pipette infusion or electroporation. For AM ester loading (Loading and Calibration of Intracellular Ion Indicators—Note 19.1), addition of the Pluronic™ F-127 (P3000MP, P6866, P6867) or PowerLoad™ (P10020) dispersing agents as well as relatively long incubation times—up to four hours—are typically necessary.

Find Fluorescent Indicators for Na+ and K+
We offer a number of fluorescent indicators for measuring Na+ and K+. Review Fluorescent Na+ and K+ Indicators—Section 21.1 in the Molecular Probes™ Handbook for more information on these products.

For Research Use. Not for human or animal therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Fluorescent
For Use With (Equipment)
Fluorescence Microscope
Product Type
PBFI
Dye Type
Potassium Indicator
Shipping Condition
Room Temperature

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text