Invitrogen™

Pro-Q™ Emerald 300 Glycoprotein Gel and Blot Stain Kit

Catalog number: P21857
Have Questions?
Invitrogen™

Pro-Q™ Emerald 300 Glycoprotein Gel and Blot Stain Kit

Catalog number: P21857
Have Questions?
Catalog Number
P21857
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P218571 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Pro-Q Emerald 300 Glycoprotein Stain provides direct detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. Pro-Q Emerald glycoprotein stain reacts with periodate-oxidized carbohydrate groups, creating a bright green-fluorescent signal on glycoproteins. The staining procedure requires only three steps: fixation, oxidation, and staining—no reduction step is required. Depending on the nature and degree of glycosylation, the Pro-Q Emerald 300 stain allows the detection of as little as 1 ng of a glycoprotein per band in gels, making this stain about 50 times more sensitive than the standard periodic acid–Schiff base method using acidic fuchsin dye. Signal can be visualized using a 300 nm UV transilluminator.

Compare all glycoprotein stains ›

This stain can be combined with general protein stains (for example SYPRO Ruby) for dichromatic detection of glycoproteins and total proteins in gel and on blots.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Blot Detection, In-Gel Detection
Detection Method
Fluorescent
Label or Dye
Pro-Q Emerald 300
Product Type
Glycoprotein Staining Kit
Shelf Life
6 Months
Product Line
PRO-Q™
Shipping Condition
Room Temperature
Target Molecule
Proteins (Glycoproteins)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text