Invitrogen™

Paclitaxel, Oregon Green™ 488 Conjugate (Oregon Green™ 488 Taxol, Flutax-2)

Catalog number: P22310
Have Questions?
Invitrogen™

Paclitaxel, Oregon Green™ 488 Conjugate (Oregon Green™ 488 Taxol, Flutax-2)

Catalog number: P22310
Have Questions?
Catalog Number
P22310
Unit Size
100 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P22310100 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This Oregon Green 488 paclitaxel derivative promises to be an important probe for labeling tubulin filaments in live cells. The fluorescent label on this new probe is attached to the 7-carbon of the paclitaxel, a strategy that permits selective binding of the probe to microtubules. Researchers have used similar fluorescent paclitaxel derivatives in a variety of systems.

Visualize staining your cell without wasting your reagents, antibodies, or time with our new Stain-iT Cell Staining Simulator.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Dye Type
Oregon Green™ 488
Product Line
Oregon Green™
Shipping Condition
Room Temperature
Label Type
Classic Dyes
Product Type
Paclitaxel
SubCellular Localization
Cytoskeleton, Tubulin

Contents & Storage

Store in freezer -5°C to -30°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text