Cytochrome b5, rHuman
Pending Discontinuation
Cytochrome b5, rHuman
Cytochrome b5, rHuman
Cytochrome b5 is a membrane-bound hemoprotein that enhances the catalytic efficiency of some P450 isoforms. Our human cytochrome b5 isRead more
Have Questions?
Catalog NumberQuantity
P2252100 μg
Catalog number P2252
Price (EUR)
588,00
Each
-
Add to cart
Quantity:
100 μg
Recurring order eligible. Learn more »
Price (EUR)
588,00
Each
Add to cart
Ask our AI about this Product
Cytochrome b5 is a membrane-bound hemoprotein that enhances the catalytic efficiency of some P450 isoforms. Our human cytochrome b5 is overexpressed in E. coli and purified to homogeneity. This enzyme is provided in solution.
For Research Use Only. Not for any animal or human therapeutic or diagnostic use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity100 μg
SpeciesHuman
RecombinantRecombinant
Shipping ConditionDry Ice
Expression SystemE. coli
Research CategoryToxicology & Drug Metabolism
Unit SizeEach
Contents & Storage
Store at -80°C
Have questions about this product? Ask our AI assisted search.
Can I use HBSS (with calcium and magnesium) to wash my cells before dissociation?
Do you offer media without phenol red (phenol red-free media)?
Is a precipitate in HBSS (with calcium and magnesium) normal?
Do I need to add additional HEPES to Hank's Balanced Salt Solution (HBSS) (Cat. No. 14025xxx)?
Can I store Hank's Balanced Salt Solution (HBSS) in the refrigerator?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Customers who viewed this item also viewed
Documents & Downloads
Certificates
Lot #Certificate TypeDateCatalog Number(s)
2494806BCertificate of AnalysisAug 17, 2022P2252
2494806ACertificate of AnalysisJun 27, 2022P2252
1875811CCertificate of AnalysisOct 09, 2021P2252
1875811BCertificate of AnalysisSep 22, 2020P2252
1875811ACertificate of AnalysisSep 09, 2020P2252
5 results displayed, search above for a specific certificate
Scientific Resources
Other products to consider
Share catalog number, name or link
1x1 image pixel for data collection