Informational:Prepare stock solutions with confidence using fresh, single-use size anhydrous DMSO (order D12345)
Invitrogen™

Pacific Blue™ C5-Maleimide

Catalog number: P30506
Have Questions?
Invitrogen™

Pacific Blue™ C5-Maleimide

Catalog number: P30506
Have Questions?
Catalog Number
P30506
Unit Size
1 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P305061 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Pacific Blue™ C5-maleimide is an excellent reagent for thiol-selective modification, quantitation and analysis. In this reaction, the thiol is added across the double bond of the maleimide to yield a thioether. This compound does not react with methionine, histidine or tyrosine. Reaction of Pacific Blue™ C5-maleimide with amines usually requires a higher pH than reaction of maleimides with thiols.

View all Pacific Blue™ dye products..

View the Fluorophore Selection Guide.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Solid
Quantity
1 mg
Product Type
Maleimide
Solubility
DMSO (Dimethylsulfoxide)
Chemical Reactivity
Thiol
Detection Method
Fluorescent
Flow Cytometer Laser Lines
405
Shipping Condition
Room Temperature
Reactive Moiety
Maleimide
Label or Dye
Pacific Blue™
Label Type
Pacific Dyes
Color
Off-White
Product Line
Molecular Probes™, Pacific Blue™
Emission
455
Excitation
410

Contents & Storage

Contains 1 vial of Pacific Blue™ dye (1 mg). Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text