Invitrogen™

Pro-Q™ Diamond Phosphoprotein Blot Stain Kit

Catalog number: P33356
Have Questions?
Invitrogen™

Pro-Q™ Diamond Phosphoprotein Blot Stain Kit

Catalog number: P33356
Have Questions?
Catalog Number
P33356
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
P333561 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Pro-Q Diamond Phosphoprotein Blot Stain Kit provides a rapid and simple method for directly detecting phosphoproteins on polyvinylidene difluoride (PVDF) or nitrocellulose membranes. The fluorescent stain binds selectively to the phosphate moiety and binding is independent of the amino acid sequence context. Because it is a direct stain, you don't need expensive antibodies or radioisotopes. The Pro-Q Diamond Phosphoprotein Blot Stain Kit can be used to analyze the native phosphorylation state of proteins obtained from tissue specimens or from body fluids.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Blot Detection
Detection Method
Fluorescent
Label or Dye
Pro-Q Diamond
Product Type
Phosphoprotein Blot Stain Kit
Shelf Life
6 Months
Product Line
PRO-Q™
Target Molecule
Proteins (Phosphoproteins)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text