Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Color | Red |
Description | pHrodo™ Red E. coli BioParticles™ Conjugate for Phagocytosis |
For Use With (Application) | Cell Analysis |
For Use With (Equipment) | Fluorescence Microscope |
Product Type | Conjugate |
Dye Type | pHrodo™ Red |
Form | Solid |
Product Line | BioParticles™, pHrodo™ |
Quantity | 5 x 2 mg |
Shipping Condition | Room Temperature |
Species | E. coli |
Unit Size | 5 x 2 mg |
Color | Deep Red |
Description | pHrodo™ Deep Red E. coli BioParticles™ Conjugate for Phagocytosis |
Detection Method | Fluorescence |
For Use With (Application) | Cell Analysis |
For Use With (Equipment) | Fluorescence Microscope, Flow Cytometer, High Content Analysis Instrument |
Product Type | Conjugate |
Dye Type | pHrodo™ Deep Red |
Form | Lyophilized Powder |
Quantity | 5 x 2 mg |
Reagent Type | Fluorogenic |
Species | E. coli |
Excitation/Emission | 640/655 |
Unit Size | 5 x 2 mg |
Color | Red |
Description | pHrodo™ Red E. coli BioParticles™ Phagocytosis Kit for Flow Cytometry |
Detection Method | Fluorescence |
For Use With (Application) | Cell Analysis |
For Use With (Equipment) | Flow Cytometer |
Product Type | Phagocytosis Kit |
Dye Type | pHrodo™ Red |
Emission | 488 nm |
Format | Bottle(s) |
Product Line | BioParticles™, pHrodo™ |
Quantity | 100 Assays |
Shipping Condition | Room Temperature |
Species | E. coli |
Unit Size | 1 kit |
Cell Type | Mammalian |
Color | Red |
Description | pHrodo™ Red S. aureus BioParticles™ Conjugate for Phagocytosis |
Detection Method | Fluorescence |
For Use With (Application) | Cell Analysis |
For Use With (Equipment) | Fluorescence Microscope |
Product Type | Conjugate |
Dye Type | pHrodo™ Red |
Form | Solid |
Product Line | BioParticles™, pHrodo™ |
Quantity | 5 x 2 mg |
Shipping Condition | Room Temperature |
Species | S. aureus |
Unit Size | 5 x 2 mg |
Color | Green |
Description | pHrodo™ Green S. aureus BioParticles™ Conjugate for Phagocytosis |
Detection Method | Fluorescence |
For Use With (Application) | Cell Analysis |
For Use With (Equipment) | Attune Acoustic Focusing Cytometer, Confocal Microscope, Floid Cell Imaging System, Fluorescence Microscope, High Content Analysis Instrument, Flow Cytometer |
Product Type | Conjugate |
Dye Type | pH-Sensitive Dyes |
Product Line | pHrodo™ |
Quantity | 5 x 2 mg |
Shipping Condition | Room Temperature |
Species | S. aureus |
Excitation/Emission | 509/533 |
Unit Size | 5 x 2 mg |
Color | Green |
Description | pHrodo™ Green Zymosan BioParticles™ Conjugate for Phagocytosis |
Detection Method | Fluorescence |
For Use With (Application) | Cell Analysis |
For Use With (Equipment) | Attune Acoustic Focusing Cytometer, Confocal Microscope, Floid Cell Imaging System, Fluorescence Microscope, High Content Analysis Instrument, Flow Cytometer |
Product Type | Conjugate |
Dye Type | pH-Sensitive Dyes |
Product Line | pHrodo™ |
Quantity | 5 x 1 mg |
Shipping Condition | Room Temperature |
Species | S. cerevisiae |
Excitation/Emission | 509/533 |
Unit Size | 5 x 1 mg |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
P35361 | 5 x 2 mg | pHrodo™ Red E. coli BioParticles™ Conjugate for Phagocytosis | 472.65 Online price 514.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P35360 | 5 x 2 mg | pHrodo™ Deep Red E. coli BioParticles™ Conjugate for Phagocytosis | 538.65 Online price 562.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10025 | 1 kit | pHrodo™ Red E. coli BioParticles™ Phagocytosis Kit for Flow Cytometry | 1,144.65 Online price 1,248.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A10010 | 5 x 2 mg | pHrodo™ Red S. aureus BioParticles™ Conjugate for Phagocytosis | 484.65 Online price 508.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P35367 | 5 x 2 mg | pHrodo™ Green S. aureus BioParticles™ Conjugate for Phagocytosis | 479.65 Online price 506.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P35365 | 5 x 1 mg | pHrodo™ Green Zymosan BioParticles™ Conjugate for Phagocytosis | 466.65 Online price 506.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|