Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Description | pHrodo iFL Red STP ester, amine reactive dye |
Molecular Weight (g/mol) | ∼1000 |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Active Ester, STP Ester |
Label or Dye | pHrodo™ Red |
Label Type | pHrodo (pH sensitive) |
Color | Red |
Product Line | pHrodo™ |
Excitation/Emission | 560/585 |
Unit Size | 1 mg |
Description | pHrodo iFL Red STP ester, amine reactive dye |
Molecular Weight (g/mol) | ∼1000 |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Active Ester, STP Ester |
Label or Dye | pHrodo™ Red |
Label Type | pHrodo (pH sensitive) |
Color | Red |
Product Line | pHrodo™ |
Excitation/Emission | 560/585 |
Unit Size | 3 x 100 μg |
Description | pHrodo iFL Green STP ester, amine reactive dye |
Molecular Weight (g/mol) | ∼1000 |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Active Ester, STP Ester |
Label or Dye | pHrodo™ Green |
Label Type | pHrodo (pH sensitive) |
Color | Green |
Product Line | pHrodo™ |
Excitation/Emission | 505/525 |
Unit Size | 1 mg |
Description | pHrodo iFL Green STP ester, amine reactive dye |
Molecular Weight (g/mol) | ∼1000 |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | Active Ester, STP Ester |
Label or Dye | pHrodo™ Green |
Label Type | pHrodo (pH sensitive) |
Color | Green |
Product Line | pHrodo™ |
Excitation/Emission | 505/525 |
Unit Size | 3 x 100 μg |
Description | pHrodo Deep Red TFP Ester |
Molecular Weight (g/mol) | ∼1300 |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | TFP ester |
Label or Dye | pHrodo™ Deep Red |
Label Type | pHrodo (pH sensitive) |
Color | Deep Red |
Product Line | pHrodo™ |
Excitation/Emission | 640/655 |
Unit Size | 3 x 100 µg |
Description | pHrodo Deep Red TFP Ester |
Molecular Weight (g/mol) | ∼1300 |
Chemical Reactivity | Amine |
Shipping Condition | Room Temperature |
Reactive Moiety | TFP ester |
Label or Dye | pHrodo™ Deep Red |
Label Type | pHrodo (pH sensitive) |
Color | Deep Red |
Product Line | pHrodo™ |
Excitation/Emission | 640/655 |
Unit Size | 1 mg |
Catalog Number | Specifications | Unit Size | Color | Excitation/Emission | Reactive Moiety | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
P36010 | 1 mg | Red | 560/585 | Active Ester, STP Ester | 678.65 Online price 708.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P36011 | 3 x 100 μg | Red | 560/585 | Active Ester, STP Ester | 395.65 Online price 401.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P36012 | 1 mg | Green | 505/525 | Active Ester, STP Ester | 681.65 Online price 694.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P36013 | 3 x 100 μg | Green | 505/525 | Active Ester, STP Ester | 393.65 Online price 402.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P35358 | 3 x 100 µg | Deep Red | 640/655 | TFP ester | 430.65 Online price 437.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
P35359 | 1 mg | Deep Red | 640/655 | TFP ester | 672.65 Online price 720.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|