ProLong™ Live Antifade Reagent, for live cell imaging
ProLong™ Live Antifade Reagent, for live cell imaging
ProLong™ Live Antifade Reagent, for live cell imaging
ProLong™ Live Antifade Reagent, for live cell imaging
Invitrogen™

ProLong™ Live Antifade Reagent, for live cell imaging

ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through useRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
P369745 x 1 mL
P369751 x 1 mL
Catalog number P36974
Price (USD)
246.00
Special offer
Online exclusive
Ends: 27-Mar-2026
300.00
Save 54.00 (18%)
Each
Add to cart
Quantity:
5 x 1 mL
Price (USD)
246.00
Special offer
Online exclusive
Ends: 27-Mar-2026
300.00
Save 54.00 (18%)
Each
Add to cart

ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through use of enzymes from the plasma membrane of E. coli. These enzymes metabolize the elements that can cause photobleaching. When dyes are illuminated, they can degrade which reduces the sample’s fluorescent intensity and leads to the creation of free radical singlet oxygen which degrades neighboring dye molecules. ProLong Live Antifade Reagent metabolizes this element, increasing the signal intensity and duration while keeping the background signal low. ProLong Live reagent has little-to-no effect on cellular viability, proliferation, or other cellular functions in experiments up to 48 hours. ProLong Live reagent provides photobleach protection in live cell experiments for fluorophores such as GFP, RFP, Hoechst 33342, MitoTracker™, LysoTracker™, and CellTracker™dyes. ProLong Live Antifade Reagent can be added to any cell culture media or buffer that is suitable for fluorescent imaging.

Specifications
Quantity5 x 1 mL
Shipping ConditionApproved for shipment at Room Temperature or on Wet Ice
Product LineProLong
Product TypeLive Antifade Reagent
Reagent TypeCell Imaging Reagent
Volume (Metric)1 mL
Unit SizeEach
Contents & Storage
Provided in 5 vials, with 1 mL each. Enough for 5000 assays (0.1 mL imaging solution/assay), or 1000 assays (0.5 mL imaging solution/assay). When stored at ≤-20°C product is stable for at least 6 months, with up to 4 freeze-thaw cycles. When stored at 2–8°C product should be used within 30 days (a precipitant may form at this temperature).

Frequently asked questions (FAQs)

After using ProLong Live Antifade Reagent on live samples and then fixing and permeabilizing, may the samples be mounted in ProLong Gold or Diamond Anifade Mountant?

Yes, this is possible.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

After using ProLong Live Antifade Reagent on my live sample, may I fix and permeabilize the sample?

Yes. We recommend washing off the ProLong Live Antifade Reagent with pre-warmed buffer or media and proceeding to your standard fixation and permeabilization steps.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

May I use ProLong Live Antifade Reagent on a fixed/permeabilized sample?

This is possible for immediate viewing, but the sample cannot be archived with ProLong Live Antifade Reagent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I want to mount my dye-labeled cells in an antifade mounting medium to keep the dyes from photobleaching. Which mounting medium do you recommend?

As dyes are illuminated for imaging, they will fade, or “photobleach”, leading to unwanted dimming and lower detection efficiency over time. An antifade mounting medium can greatly reduce photobleaching. If you wish to label live cells, use of ProLong Live Antifade Reagent is helpful. If you wish to mount fixed cells after labeling, and then image immediately and then discard, SlowFade Diamond Antifade Mountant stays liquid but has good refractive index. If you wish to mount your sample and then archive the slides, ProLong Diamond Antifade Mountant will harden to a better refractive index and allow for archiving of the sample for up to several weeks, or even months. Unlike other antifade mounting media, these work well with fluorescent proteins for immediate viewing (archiving fluorescent proteins is not possible), and they are packaged with or without DAPI. More information on these can be found here (https://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/fluorescence-microscopy-and-immunofluorescence-if/mounting-medium-antifades/prolong-gold-antifade.html).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

My fluorescent dye signal is fading as I image it. What can I do to stop this?

All fluorescent dyes will fade, or “photobleach”, to at least some extent when exposed to strong light at the wavelengths they absorb. Here are some causes for photobleaching and ways to fix the problem:

1) Cause of photobleacing - Generation of free radicals and singlet oxygen
Remedy - i) Use an antifade reagent, which has antioxidants and free radical scavengers:

ii) For live-cell imaging of fluorescent dyes and proteins, we recommend ProLong Live Antifade Reagent which can be added to the cell media or buffer. ProLong Live Antifade Reagent can significantly increase the stability over time for reagents as well as fluorescent proteins, like GFP, without affecting cell health, for up to 24 hours.

iii) For immediate analysis and short-term storage of fixed samples, we recommend SlowFade Diamond Antifade Mountant (it is a liquid mountant and can be used for immediate viewing and then disposal of the sample within a day).

iv) For long-term analysis of Alexa Fluor dyes in fixed samples, we recommend a mountant that hardens, such as ProLong Diamond Antifade Mountant. The harrdening of the mountant also slows diffusion of free radicals).

v) For long-term analysis of all dyes and fluorescent proteins in fixed samples, we recommend ProLong Diamond Antifade Mountant, suitable for archiving slides.

2) Cause of photobleaching - Dye is particularly sensitive to structural modification upon exposure to light.
Remedy - i) Choose a more photostable dye, such as many of our Alexa Fluor dyes.

3) Cause of photobleaching - Intense Illlumination
Remedy - i) Reduce light exposure, for example by reducing laser power or using neutral density filters.

ii) Minimize the viewing time of labeled sample, and close shutter when not viewing.

iii) Use an objective with a lower numerical aperture, such as a lower-power objective.

You can find more information on choosing an antifade reagent on the link below http://www.thermofisher.com/us/en/home/life-science/cell-analysis/cellular-imaging/fluorescence-microscopy-and-immunofluorescence-if/mounting-medium-antifades.html.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (15)

Citations & References
Abstract
Persistent Replication of a Chikungunya Virus Replicon in Human Cells Is Associated with Presence of Stable Cytoplasmic Granules Containing Nonstructural Protein 3.
Authors:Remenyi R, Gao Y, Hughes RE, Curd A, Zothner C, Peckham M, Merits A, Harris M,
Journal:J Virol
PubMed ID:29875241
'Chikungunya virus (CHIKV), a mosquito-borne human pathogen, causes a disabling disease characterized by severe joint pain that can persist for weeks, months, or even years in patients. The nonstructural protein 3 (nsP3) plays essential roles during acute infection, but little is known about the function of nsP3 during chronic disease. ... More
Diatom bloom-derived biotoxins cause aberrant development and gene expression in the appendicularian chordate Oikopleura dioica.
Authors:Torres-Águila NP, Martí-Solans J, Ferrández-Roldán A, Almazán A, Roncalli V, D'Aniello S, Romano G, Palumbo A, Albalat R, Cañestro C,
Journal:Commun Biol
PubMed ID:30272001
'Investigating environmental hazards than could affect appendicularians is of prime ecological interest because they are among the most abundant components of the mesozooplankton. This work shows that embryo development of the appendicularian Oikopleura dioica is compromised by diatom bloom-derived biotoxins, even at concentrations in the same range as those measured ... More
Complex bile duct network formation within liver decellularized extracellular matrix hydrogels.
Authors:Lewis PL, Su J, Yan M, Meng F, Glaser SS, Alpini GD, Green RM, Sosa-Pineda B, Shah RN,
Journal:Sci Rep
PubMed ID:30111800
'The biliary tree is an essential component of transplantable human liver tissue. Despite recent advances in liver tissue engineering, attempts at re-creating the intrahepatic biliary tree have not progressed significantly. The finer branches of the biliary tree are structurally and functionally complex and heterogeneous and require harnessing innate developmental processes ... More
Exopolysaccharide production in Caulobacter crescentus: A resource allocation trade-off between protection and proliferation.
Authors:Herr KL, Carey AM, Heckman TI, Chávez JL, Johnson CN, Harvey E, Gamroth WA, Wulfing BS, Van Kessel RA, Marks ME,
Journal:PLoS One
PubMed ID:29293585
'Complex and interacting selective pressures can produce bacterial communities with a range of phenotypes. One measure of bacterial success is the ability of cells or populations to proliferate while avoiding lytic phage infection. Resistance against bacteriophage infection can occur in the form of a metabolically expensive exopolysaccharide capsule. Here, we ... More
Mmf1p Couples Amino Acid Metabolism to Mitochondrial DNA Maintenance in Saccharomyces cerevisiae.
Authors:Ernst DC, Downs DM,
Journal:MBio
PubMed ID:29487232
'A variety of metabolic deficiencies and human diseases arise from the disruption of mitochondrial enzymes and/or loss of mitochondrial DNA. Mounting evidence shows that eukaryotes have conserved enzymes that prevent the accumulation of reactive metabolites that cause stress inside the mitochondrion. 2-Aminoacrylate is a reactive enamine generated by pyridoxal 5''-phosphate-dependent ... More