Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: PEVRKLPEEK HIPEELKPTG KELPDSQLVQ PLVYMERLEL IRNVCRDDAL KNLSHTPVSK FVLDRIFVCD KHKILFCQTP KVGNTQWKKV LIVLNGAFSS IEEIPENVVH DHEKNGLPRL SSFSDAEIQK RL
Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
This protein encoded by this gene transfers sulfate to the C-3 hydroxyl of terminal glucuronic acid of protein- and lipid-linked oligosaccharides. This protein was first identified as a sulfotransferase that acts on the human natural killer-1 (HNK-1) glycan; HNK-1 is a carbohydrate involved in neurodevelopment and synaptic plasticity.[provided by RefSeq, Feb 2011].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carbohydrate sulfotransferase 10; HNK-1 sulfotransferase; HNK-1ST; huHNK-1ST
Gene Aliases: CHST10; HNK-1ST; HNK1ST
UniProt ID: (Human) O43529
Entrez Gene ID: (Human) 9486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support