Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: GLNVIKEAEA QLWWAAKELR RTKKLSDYVG KNEKTKIIAK IQQRGQGAPA REPIISSEEQ KQLMLYYHRR QEELKRLEEN DDDAYLNSPW ADNTALKR
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The function of the C21orf59 protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cilia- and flagella-associated protein 298; prostate cancer upregulated protein 1; Protein kurly homolog
Gene Aliases: C21orf48; C21orf59; CFAP298; CILD26; FBB18; Kur
UniProt ID: (Human) P57076
Entrez Gene ID: (Human) 56683
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support