Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: AQDSTQASTP GSPLSPTEYE RFFALLTPTW KAETTCRLRA THGCRNPTLV QLDQYENHGL VPDGAVCSNL PYASWFESFC QFTHYRCS
Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The protein encoded by this gene is similar to proacrosin binding protein sp32 precursor found in mouse, guinea pig, and pig. This protein is located in the sperm acrosome and is thought to function as a binding protein to proacrosin for packaging and condensation of the acrosin zymogen in the acrosomal matrix. This protein is a member of the cancer/testis family of antigens and it is found to be immunogenic. In normal tissues, this mRNA is expressed only in testis, whereas it is detected in a range of different tumor types such as bladder, breast, lung, liver, and colon. [provided by RefSeq, Jul 2008].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACRBP; Acrosin-binding protein; Acrosin-binding protein, 60 kDa form; Cancer/testis antigen 23; Cancer/testis antigen OY-TES-1; CT23; proacrosin binding protein sp32; Proacrosin-binding protein sp32; testicular tissue protein Li 10
Gene Aliases: ACRBP; CT23; OY-TES-1; SP32
UniProt ID: (Human) Q8NEB7
Entrez Gene ID: (Human) 84519
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support