Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: NLVVINKPYG LPVHGGPGVQ LCITDVLPIL AKMLHGHKAE PLHLCHRLDK ETTGVMVLAW DKDMAHQVQE LFRTRQVVKK YWAITVHVP
Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pseudouridylate synthase RPUSD4, mitochondrial; RNA pseudouridylate synthase domain-containing protein 4
Gene Aliases: RPUSD4
UniProt ID: (Human) Q96CM3
Entrez Gene ID: (Human) 84881
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support