Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LTADVQVFPR PEPFVVDEEI DPIPKVINTD LEIVGFIDIA DISSPPVLSR HLVLPIALNK EGDEVGTGIT DDNEDENSAN QIAGKIPNFC VLLHGSLKVE GMVAIVQL
Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
C15orf44 is an uncharacterized protein that bears a P-loop nucleoside triphosphate hydrolase (NTPase) domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Integrator complex subunit 14; UPF0464 protein C15orf44; UPF0464 protein C15orf44 homolog; von Willebrand factor A domain-containing protein 9
Gene Aliases: 2010321M09Rik; C15orf44; INTS14; RGD1359616; VWA9
UniProt ID: (Human) Q96SY0, (Rat) Q66H58, (Mouse) Q8R3P6
Entrez Gene ID: (Human) 81556, (Rat) 300782, (Mouse) 69882
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support