Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: FLPYYICVADE DDYRDFVNLD HAHSLLRDYQ QREGIAMDQL LSQSLPNDGD EKYEKTIIKS GDQTFYKFMK RIAACQEQIL
Highest antigen sequence identity to the following orthologs - mouse 72%, rat 68%.
Over expression of PDCD2L attenutates TNF-alpha release in Daudi cells. PDCD2L over-expression restrained proliferation of HEK293T cells. DNA/flow cytometry analysis showed that the over-expression of PDCD2L severely delays cell cycle progression at S phase. These all suggest that PDCD2L might play a role in apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC13096; programmed cell death 2-like; Programmed cell death protein 2-like
Gene Aliases: PDCD2L
UniProt ID: (Human) Q9BRP1
Entrez Gene ID: (Human) 84306
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support