Search
Search
Invitrogen
Promotions
View available promotion(s)
Promo Code: RGLOW25 Save up to $900 and get the limited-edition holiday sweatshirt + matching beanie It’s been a year – this is your badge of honor for fluorescing through it all. Offer available until December 27.. Learn more
Spend Now, Save Big Later! Spend Now, Save Big Later! Make your New year's resolution to save: earn up to $3,500 off a future January offer when you purchase a qualifying amount before December 31st. Learn more
Immunogen sequence: GICAERTAIQ KAVSEGYKDF RAIAIASDMQ DDFISPCGAC RQVMREFGTN WPVYMTKPDG TYIVMTVQE
Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 87%.
This gene encodes an enzyme involved in pyrimidine salvaging. The encoded protein forms a homotetramer that catalyzes the irreversible hydrolytic deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. It is one of several deaminases responsible for maintaining the cellular pyrimidine pool. Mutations in this gene are associated with decreased sensitivity to the cytosine nucleoside analogue cytosine arabinoside used in the treatment of certain childhood leukemias.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytidine aminohydrolase; Cytidine deaminase; cytosine nucleoside deaminase; EC 3.5.4.5; small cytidine deaminase
Gene Aliases: CDA; CDD
UniProt ID: (Human) P32320
Entrez Gene ID: (Human) 978
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support