Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Percent identity with other species by BLAST analysis: Human, Gorilla (100%) Monkey (88%) Marmoset, Rat, Elephant, Pig, Bloom alga (82%).
The neuropeptide galanin modulates a variety of physiologic processes including cognition/memory, sensory/pain processing, hormone secretion, and feeding behavior. The human galanin receptors are G protein-coupled receptors that functionally couple to their intracellular effector through distinct signaling pathways. GALR3 is found in many tissues and may be expressed as 1.4-, 2.4-, and 5-kb transcripts. GALR3 has been reported in brain, testis, adrenal gland, and pancreas. An EST has been isolated from an ovarian thecoma fibroma library.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gal receptor-3; Gal-r3; Gal3 receptor; GAL3-R; Galanin r3 receptor; Galanin receptor type 3; galanin receptor, family member 3; GALR-3; OTTHUMP00000028820
Gene Aliases: GALNR3; GALR3
UniProt ID: (Human) O60755, (Rat) O88626
Entrez Gene ID: (Human) 8484, (Rat) 29235
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support