Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Positive Control: 293T, A431, HeLa, HepG2
Predicted Reactivity: Rat (99%), Xenopus laevis (95%), Dog (100%), Rabbit (97%), Sheep (99%), Bovine (98%)
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAB family members are small, RAS-related GTP-binding proteins that are important regulators of vesicular transport. Each RAB protein targets multiple proteins that act in exocytic / endocytic pathways. This gene encodes a RAB family member that regulates vesicle traffic in the late endosomes and also from late endosomes to lysosomes. This encoded protein is also involved in the cellular vacuolation of the VacA cytotoxin of Helicobacter pylori. Mutations at highly conserved amino acid residues in this gene have caused some forms of Charcot-Marie-Tooth (CMT) type 2 neuropathies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: rab-7; RAB7, member RAS oncogene family; Ras-associated protein RAB7; Ras-related protein Rab-7a
Gene Aliases: PRO2706; RAB7; RAB7A
UniProt ID: (Human) P51149, (Mouse) P51150
Entrez Gene ID: (Human) 7879, (Mouse) 19349
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support