Invitrogen™

Power Blotter Cassette XL

Catalog number: PB0003
Have Questions?
Invitrogen™

Power Blotter Cassette XL

Catalog number: PB0003
Have Questions?
Catalog Number
PB0003
Unit Size
1 cassette
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PB00031 cassette
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Invitrogen Power Blotter Cassette XL is part of the Power Blotter System designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter Cassette XL is the larger version of the standard-size Power Blotter Cassette, and can simultaneously transfer up to four mini-size or two midi-size gels. This item is made available as a replacement or additional cassette for the Power Blotter Station.

The Power Blotter Cassette XL can be used with Power Blotter Select Transfer Stacks for rapid transfers, the Power Blotter 1-Step Transfer Buffer for semi-dry transfers, or alternatively with traditional buffers for longer semi-dry transfers.

Learn more about the Power Blotter System ›

Related products
Power Blotter Cassette

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Western Blot
For Use With (Equipment)
PB0010 Power Blotter Station
Membrane Compatibility
Nitrocellulose, PVDF
Mode of Transfer
Semi-Dry
Product Line
Power Blotter
Throughput
2 Midi or 4 Mini Gels
Type
Blotter Cassette
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text