Invitrogen™

Power Blotter System

Catalog number: PB0012
Have Questions?
Invitrogen™

Power Blotter System

Catalog number: PB0012
Have Questions?
Catalog Number
PB0012
Unit Size
1 system
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PB00121 system
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Invitrogen Power Blotter System is designed specifically for rapid semi-dry transfer of proteins from polyacrylamide gels to nitrocellulose or PVDF membranes in 5 to 12 minutes. The Power Blotter features an integrated power supply optimized to enable consistent and high-efficiency protein transfer.

Features of the Power Blotter System include:
Efficient—high transfer efficiency with a broad range of protein sizes compared to conventional semi-dry or wet (tank) transfer methods
Fast—5 to 12 minute transfer when used with Power Blotter Select Transfer Stacks or Power Blotter 1-Step Transfer Buffer
Integrated power supply—seamless operation between control unit and cassette; no need for external power supply
Easy-touch programming—convenient pre-programmed transfer methods or create, save, and run customized transfer methods
Flexible—simultaneous transfer of 1 to 2 mini-size or 1 midi-size gel using the Power Blotter Cassette
Versatile—compatible with Power Blotter Select Transfer Stacks and Pierce 1-Step Transfer Buffer for rapid blotting programs or Towbin transfer buffer for conventional semi-dry transfer methods

Learn more about the semi-dry transfer device: Power Blotter System

The Power Blotter Cassette, included in the Power Blotter System, enables the simultaneous transfer of up to two mini-size or one midi-size gel. The included Power Blotter Station has an easy-to-use color LCD touchscreen interface with pre-programmed transfer methods for different numbers and sizes of gels and ranges of molecular weight protein. The easy-touch programming feature allows custom transfer settings to be quickly created, saved, and run.

The Power Blotter System achieves highly efficient transfer in seven minutes when used with Power Blotter Select Transfer Stacks or ten minutes with Power Blotter 1-Step Transfer Buffer. The instrument is also effective for standard 30- to 60-minute semi-dry transfer protocols based on traditional buffers.

Contents of the Power Blotter System:
Power Blotter Station
Power Blotter Cassette

Related products
Power Blotter XL System

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Mode of Transfer
Semi-Dry
Product Line
Power Blotter
Throughput
1 Midi or 2 Mini Gels
Type
Blotter System
Shipping Condition
Room Temperature

Contents & Storage

• Power Blotter Station
• Power Blotter Cassette

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text