Invitrogen™

Power Blotter Select Transfer Stacks, PVDF, mini

Catalog number: PB5240
Have Questions?
Invitrogen™

Power Blotter Select Transfer Stacks, PVDF, mini

Catalog number: PB5240
Have Questions?
Catalog Number
PB5240
Unit Size
40 stacks
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
10 stacks
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
PB524040 stacks
-
PB521010 stacks
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Invitrogen Power Blotter Select Transfer Stacks are used to transfer proteins using the Power Blotter System. They are disposable, pre-assembled stacks that come with an integrated PVDF transfer membrane for dry blotting of proteins from polyacrylamide gels. Each Power Blotter Select Transfer Stack contains filter paper, membrane, and the appropriate cathode and anode buffers incorporated into a solid gel matrix to allow fast, reliable transfer of proteins.

Features of the Power Blotter Select Transfer Stacks:
Rapid transfer—the ready-to-use transfer stack takes little setup time and protein transfers take place in as little as seven minutes
Efficient and even transfer—stacks are made with high-quality membranes that provide high binding efficiency, uniform transfer, and high detection sensitivity
Increased reproducibility—preassembled stacks provide consistent performance

Learn more about the use of the Power Blotter Select Transfer Stacks in Power Blotter System

PVDF membrane
The PVDF membrane (0.2 μm pore size, low fluorescence) in the stack has higher binding capacity than nitrocellulose. It is pre-activated and ready for use without any pretreatment with alcohols. The membrane is compatible with commonly used detection methods such as staining, immunodetection, fluorescence, and radiolabeling. The proteins bind to the membrane through hydrophobic interactions. The protein-binding capacity is 240 μg/cm2.

Mini transfer stacks
The mini-size (8 cm x 8 cm) Power Blotter Select Stacks can be used with all Invitrogen Novex, Bolt, and NuPAGE mini-size gels or equivalent.

Related products
Power Blotter Select Transfer Stacks, PVDF, regular size
Power Blotter Select Transfer Stacks, nitrocellulose, mini size
Power Blotter Select Transfer Stacks, nitrocellulose, regular size

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Length (Metric)
8 cm
Width (Metric)
8 cm
Format
Transfer Stack
Material
PVDF
Pore Size
0.2 μm
Shipping Condition
Room Temperature
Dimensions (LxW)
8 cm x 8 cm
Product Line
Power Blotter
Sufficient For
40 Blots

Contents & Storage

40 mini-size transfer stacks (8 cm x 8 cm)

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text